T5376
Glyceryl triacetate
≥99%
Synonym(s):
1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glycerol triacetate, Triacetin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3COOCH2)2CHOCOCH3
CAS Number:
Molecular Weight:
218.20
Beilstein:
1792353
EC Number:
MDL number:
UNSPSC Code:
41141833
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥99%
bp
258-260 °C (lit.)
density
1.16 g/mL at 25 °C (lit.)
lipid type
neutral glycerides
SMILES string
CC(=O)OCC(COC(C)=O)OC(C)=O
InChI
1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3
InChI key
URAYPUMNDPQOKB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
298.4 °F - closed cup
Flash Point(C)
148 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Boyong Wan et al.
Analytica chimica acta, 681(1-2), 63-70 (2010-11-03)
Wavelet analysis is developed as a preprocessing tool for use in removing background information from near-infrared (near-IR) single-beam spectra before the construction of multivariate calibration models. Three data sets collected with three different near-IR spectrometers are investigated that involve the
G Sharma et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 76(2), 159-169 (2010-07-27)
Insulin loaded microemulsions were developed adopting a low shear reverse micellar approach using didoceyldimethylammonium bromide (DMAB) as the surfactant, propylene glycol (PG) as the co-surfactant, triacetin (TA) as the oil phase and insulin solution as the aqueous phase. A ternary
C N Madhavarao et al.
Journal of inherited metabolic disease, 32(5), 640-640 (2009-08-18)
Canavan disease (CD) is a fatal dysmyelinating genetic disorder associated with aspartoacylase deficiency, resulting in decreased brain acetate levels and reduced myelin lipid synthesis in the developing brain. Here we tested tolerability of a potent acetate precursor, glyceryl triacetate (GTA)
Shanhui Zhu et al.
Bioresource technology, 130, 45-51 (2013-01-12)
The synthesis of bioadditives for biofuels from glycerol esterification with acetic acid was performed over zirconia supported heteropolyacids catalysts using H(4)SiW(12)O(40) (HSiW), H(3)PW(12)O(40) (HPW) and H(3)PMo(12)O(40) (HPMo) as active compounds. The as-prepared catalysts were characterized by N(2)-physisorption, XRD, Raman spectroscopy
Huayin Pu et al.
Journal of agricultural and food chemistry, 59(10), 5738-5745 (2011-04-26)
An oral colon-targeting controlled release system based on resistant starch acetate (RSA) as a film-coating material was developed. The RSA was successfully synthesized, and its digestion resistibility could be improved by increasing the degree of substitution (DS), which was favorable
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service