Skip to Content
Merck
All Photos(1)

Key Documents

T4816

Sigma-Aldrich

Clear-view Snap-Cap microtubes

capacity 1.5 mL, natural, low retention

Synonym(s):

low retention microtube, snap-cap vial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.22

material

conical bottom
flat cap
polypropylene

sterility

non-sterile

feature

RNase and DNase free
neck thread Push fit
low retention

packaging

pack of 250 ea

manufacturer/tradename

Excel Scientific MCTS 1.5N

parameter

-20,000 × g max. RCF

technique(s)

PCR: suitable

capacity

1.5 mL

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

  • Superior clarity for sample viewing
  • A consistent, ring-lock seal prevents inadvertent opening
  • Angled cap lid makes opening easy without thumb fatigue
  • Large flat tops for ease of identification and a pierceable cap
  • Large, crisp reference lines and graduation numbers
  • Certified RNase/DNase free

Application

Clear-view Snap-Cap microtubes have been used in iCLIP (individual-nucleotide resolution UV crosslinking and immunoprecipitation) protocol. They have been used for the preparation of serum samples for MS/MS (mass spectrometry) analysis.

Legal Information

Clear-view is a trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Individual-nucleotide-resolution UV Cross-linking and Immunoprecipitation (iCLIP) of UPF1.
Zund D and Muhlemann O
Bio-protocol (2014)
Ruben N Pinto et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 95(9), 976-984 (2019-07-12)
Deleterious changes, collectively termed as storage lesions, alter the characteristics of red blood cell (RBC) morphology during in vitro storage. Due to gradual loss of cellular membrane, RBCs lose their original biconcave disk shape and begin a process of spherical
Pre-analytical-related variability influencing serum peptide profiles demonstrated in a mass spectrometry-based search for colorectal and prostate cancer biomarkers.
Karczmarski J, et al.
Acta Biochimica Polonica, 60, 417-425 (2013)
Minh Doan et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(35), 21381-21390 (2020-08-26)
Stored red blood cells (RBCs) are needed for life-saving blood transfusions, but they undergo continuous degradation. RBC storage lesions are often assessed by microscopic examination or biochemical and biophysical assays, which are complex, time-consuming, and destructive to fragile cells. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service