SRP5014
CK1 δ, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
CK1D, CSNK1D, HCKID
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
70-94 nmol/min·mg
mol wt
~72 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... CSNK1D(1453)
General description
Casein kinase 1 δ (CK1δ) is a member of the CK1 family of serine/threonine protein kinases which play an important role in diverse cell processes, including DNA replication and repair.
Biochem/physiol Actions
CK1δ is a regulator of Yes-associated protein (YAP) transcription coactivator, which is a key regulator of organ size and a candidate human oncogene. CK1d is activated by CCK2R and it then phosphorylates polycystin 2 (PKD2) at Ser244. Phosphorylation of PKD2 leads to its nuclear accumulation and efficient phosphorylation of nuclear PKD2 substrates in human gastric cancer cells. CKId can phosphorylate in vitro deoxycytidine kinase (dCK), which is a key enzyme in the salvage of deoxynucleosides. Phosphorylation of dCK by CKId in vitro correlates with increased activity of this enzyme.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Phosphorylation at Ser244 by CK1 determines nuclear localization and substrate targeting of PKD2
J. von Blume
The Embo Journal (2007)
A coordinated phosphorylation by Lats and CK1 regulates YAP stability through SCF?-TRCP
Bin Zhao
Genes & Development (2010)
The CK1 Family: Contribution to Cellular Stress Response and Its Role in Carcinogenesis
Uwe Knippschild
Frontiers in Oncology (2014)
Caroline Smal et al.
Archives of biochemistry and biophysics, 502(1), 44-52 (2010-07-20)
Deoxycytidine kinase (dCK) is a key enzyme in the salvage of deoxynucleosides and in the activation of several anticancer and antiviral nucleoside analogues. We recently showed that dCK was activated in vivo by phosphorylation of Ser-74. However, the protein kinase
J von Blume et al.
The EMBO journal, 26(22), 4619-4633 (2007-10-27)
Protein kinase D2 (PKD2), a member of the PKD family of serine/threonine kinases, is localized in various subcellular compartments including the nucleus where the kinase accumulates upon activation of G-protein-coupled receptors. We define three critical post-translational modifications required for nuclear
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service