Skip to Content
Merck
All Photos(2)

Key Documents

SML3797

Sigma-Aldrich

Biotin-HPDP

≥95% (HPLC)

Synonym(s):

(3aS,4S,6aR)-Hexahydro-2-oxo-N-[6-[[1-oxo-3-(2-pyridinyldithio)propyl]amino]hexyl]-1H-thieno[3,4-d]imidazole-4-pentanamide, HPDP-Biotin, N-[6-(Biotinamido)hexyl]-3′-(2′-pyridyldithio)propionamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H37N5O3S3
CAS Number:
Molecular Weight:
539.78
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

S1[C@H]([C@H]3NC(=O)N[C@H]3C1)CCCCC(=O)NCCCCCCNC(=O)CCSSc2ncccc2

InChI

1S/C24H37N5O3S3/c30-20(10-4-3-9-19-23-18(17-33-19)28-24(32)29-23)25-13-6-1-2-7-14-26-21(31)12-16-34-35-22-11-5-8-15-27-22/h5,8,11,15,18-19,23H,1-4,6-7,9-10,12-14,16-17H2,(H,25,30)(H,26,31)(H2,28,29,32)/t18-,19-,23-/m0/s1

InChI key

QLPHBNRMJLFRGO-YDHSSHFGSA-N

Biochem/physiol Actions

Biotin-HPDP (HPDP-Biotin) is a membrane-permeable and reversible biotinylating reagent that is used to biotinylate target proteins via covalent disulfide bond (-S-S-) formation with free sulfhydryl (-SH) groups. The biotin-tagged proteins can then be subjected to affinity labeling or isolation/purification using avidin conjugates or beads. Unlike tranditonal biotinylation methods, Biotin-HPDP tag can be removed with reducing agents such as dithiothreitol (DTT) without the need of harsh reagents to dissociate the high affinity interaction between avidin and biotin.
Membrane-permeable, reversible biotinylating reagent for affinity labeling and isolation of proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-Background Acyl-Biotinyl Exchange Largely Eliminates the Coisolation of Non- S-Acylated Proteins and Enables Deep S-Acylproteomic Analysis
Analytical Chemistry, 91(15), 9858-9866 (2019)
Copper dependence of the biotin switch assay: modified assay for measuring cellular and blood nitrosated proteins
Free Radical Biology & Medicine, 44(7), 1362-1372 (2008)
Localization and partial characterization of the oligomeric disulfide-linked molecular weight 95,000 protein (triadin) which binds the ryanodine and dihydropyridine receptors in skeletal muscle triadic vesicles
Biochemistry, 30(30), 7507-7513 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service