SML3554
6-(Methylsulfinyl)hexyl isothiocyanate
≥98% (HPLC)
Synonym(s):
1-Isothiocyanato-6-(methylsulfinyl)hexane, 6-MITC, 6-MSITC, Hesperin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
oil
color
colorless to yellow
storage temp.
2-8°C
SMILES string
O=S(C)CCCCCCN=C=S
InChI
1S/C8H15NOS2/c1-12(10)7-5-3-2-4-6-9-8-11/h2-7H2,1H3
InChI key
XQZVZULJKVALRI-UHFFFAOYSA-N
Biochem/physiol Actions
6-(Methylsulfinyl)hexyl isothiocyanate (6-MITC) is a major bioactive compound in wasabi that exhibits antimicrobial, anti-inflammatory, antiplatelet, antioxidant and anticancer properties (mean log GI50 = −5.41 (3.9 μM) in a human cancer cell panel). 6-MITC concurrently induces autophagy and mitotic arrest, suppresses NF-κβ signaling by competitive inhibition of GSK-3β.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Antimutagenic and Chemopreventive Properties of 6-(Methylsulfinyl) Hexyl Isothiocyanate on TK6 Human Cells by Flow Cytometry
Frontiers in Pharmacology, 11 (2020)
6-(methylsulfinyl)hexyl isothiocyanate (6-MITC) from Wasabia japonica alleviates inflammatory bowel disease (IBD) by potential inhibition of glycogen synthase kinase 3 beta (GSK-3?)
European Journal of Medicinal Chemistry, 216 (2021)
Molecular and cellular biology, 40(6) (2020-01-15)
Nrf2 (NF-E2-related-factor 2) is a stress-responsive transcription factor that protects cells against oxidative stresses. To clarify whether Nrf2 prevents Alzheimer's disease (AD), AD model AppNL-G-F/NL-G-F knock-in (AppNLGF ) mice were studied in combination with genetic Nrf2 induction model Keap1FA/FA mice.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service