Skip to Content
Merck
All Photos(1)

Key Documents

SML3511

Sigma-Aldrich

S-217622

≥98% (HPLC)

Synonym(s):

(6E)-6-[(6-chloro-2-methyl-2H-indazol-5-yl)imino]dihydro-3-[(1-methyl-1H-1,2,4-triazol-3-yl)methyl]-1-[(2,4,5-trifluorophenyl)methyl]-1,3,5-Triazine-2,4(1H,3H)-dione, Ensitrelvir, S 217622

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17ClF3N9O2
CAS Number:
Molecular Weight:
531.88
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N/C(N(C(N1CC2=NN(C=N2)C)=O)CC3=CC(F)=C(F)C=C3F)=N\C4=CC5=CN(N=C5C=C4Cl)C

InChI

1S/C22H17ClF3N9O2/c1-32-7-12-4-18(13(23)5-17(12)30-32)28-20-29-21(36)35(9-19-27-10-33(2)31-19)22(37)34(20)8-11-3-15(25)16(26)6-14(11)24/h3-7,10H,8-9H2,1-2H3,(H,28,29,36)

InChI key

QMPBBNUOBOFBFS-UHFFFAOYSA-N

Biochem/physiol Actions

S-217622 is an orally available potent and selective noncovalent and nonpeptidic inhibitor of SARS-CoV-2 3C-like protease (3CLpro, or Mpro) that exhibits antiviral activity in vitro against varies variants of SARS-CoV-2 and other coronaviruses. S-217622 potently inhibits intrapulmonary replication of SARS-CoV-2 in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of S-217622, a Noncovalent Oral SARS-CoV-2 3CL Protease Inhibitor Clinical Candidate for Treating COVID-19.
Journal of medicinal chemistry, 65(9), 6499-6512 (2022)
S-217622, a 3CL Protease Inhibitor and Clinical Candidate for SARS-CoV-2
Tyndall JDA
Journal of Medicinal Chemistry, 65(9), 6496-6498 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service