Skip to Content
Merck
All Photos(1)

Key Documents

SML3409

Sigma-Aldrich

ATN-224

≥95% (elemental analysis)

Synonym(s):

2-hydroxy-N,N,N-trimethylethanaminium tetrathiomolybdate, ALXN1840, Bis-Choline-Tetrathiomolybdate, Tiomolibdate Choline, WTX101

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H28N2O2 · MoS4
CAS Number:
Molecular Weight:
432.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (elemental analysis)

form

powder

storage condition

desiccated

color

, Orange to very dark red

solubility

H2O: ≥0.1 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

S=[Mo]([S-])([S-])=S.C[N+](C)(CCO)C.C[N+](C)(CCO)C

InChI

1S/2C5H14NO.Mo.4S/c2*1-6(2,3)4-5-7;;;;;/h2*7H,4-5H2,1-3H3;;;;;/q2*+1;;;;2*-1

InChI key

NEYVHGQOGHJAAD-UHFFFAOYSA-N

Biochem/physiol Actions

ATN-224 (Bis-Choline-Tetrathiomolybdate) is an orally available and potent inhibitor of superoxide dismutase 1 (SOD1) that exhibit potent anticancer activities. ATN-224 is a copper chelator that rapidly lowers free coper levels and improve clinical symptoms in Wilson′s disease patients.
Inhibitor of superoxide dismutase 1 (SOD1); Reactive oxygen species (ROS) scavenging blocker

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service