SML3004
NMDAR/TRPM4 interface inhibitor C8 dihydrochloride
≥98% (HPLC)
Synonym(s):
C8 dihydrochloride, Compound 8 dihydrochloride, N1-[(3-Bromophenyl)methyl]-N1-ethyl-1,2-ethanediamine dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H17BrN2·2HCl
CAS Number:
Molecular Weight:
330.09
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder or solid
storage condition
desiccated
color
white to beige
solubility
H2O: 2 mg/mL, clear
storage temp.
−20°C
SMILES string
NCCN(CC)CC1=CC=CC(Br)=C1.Cl.Cl
Biochem/physiol Actions
Compound 8 (C8) is a small molecule that directly targets TRPM4 TwinF domain and blocks its interaction with GluN2A/GluN2B (NR2A/NR2B) I4 domain. C8 prevents NMDAR/TRPM4 interaction-dependent excitotoxicity (death protection EC50 = 2.1 μM 24 h post 10-min exposure of primary murine hippocampal neurons to 20 μM NMDA) without affecting, and even enhancing, NMDAR-mediated essential functions. C8 protects mice from MCAO-induced brain damage and NMDA-induced retinal ganglion cell (RGC) loss in mice in vivo (40 mg/kg i.p. at -16h, -3h before, 0h, 3h, 24h after MCAO or 20 nmol NMDA/2.0 μL by intravitreal injection).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jing Yan et al.
Science (New York, N.Y.), 370(6513) (2020-10-10)
Excitotoxicity induced by NMDA receptors (NMDARs) is thought to be intimately linked to high intracellular calcium load. Unexpectedly, NMDAR-mediated toxicity can be eliminated without affecting NMDAR-induced calcium signals. Instead, excitotoxicity requires physical coupling of NMDARs to TRPM4. This interaction is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service