Skip to Content
Merck
All Photos(1)

Key Documents

SML2144

Sigma-Aldrich

TB5

≥98% (HPLC)

Synonym(s):

(2E)-1-(5-bromothiophen-2-yl)-3-[4-(dimethylamino)phenyl]prop-2-en-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14BrNOS
CAS Number:
Molecular Weight:
336.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)C(C=C1)=CC=C1/C=C/C(C2=CC=C(Br)S2)=O

InChI

1S/C15H14BrNOS/c1-17(2)12-6-3-11(4-7-12)5-8-13(18)14-9-10-15(16)19-14/h3-10H,1-2H3/b8-5+

Biochem/physiol Actions

TB5 is a potent and selective reversible inhibitor of monoamine oxidase B (MAOB) with a Ki value of 110 nM. A reversible inhibitor should have an advantage over irreversible inhibitors such as selegiline in limiting side effects. Parallel artificial membrane permeation (PAMPA) assays indicated TB5 should be able to cross the blood–brain barrier. TB5 was found nontoxic to cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bijo Mathew et al.
ChemMedChem, 11(11), 1161-1171 (2016-05-10)
A series of (2E)-1-(5-bromothiophen-2-yl)-3-(para-substituted phenyl)prop-2-en-1-ones (TB1-TB11) was synthesized and tested for inhibitory activity toward human monoamine oxidase (hMAO). All compounds were found to be competitive, selective, and reversible toward hMAO-B except (2E)-1-(5-bromothiophen-2-yl)-3-(4-nitrophenyl)prop-2-en-1-one (TB7) and (2E)-1-(5-bromothiophen-2-yl)-3-(4-chlorophenyl)prop-2-en-1-one (TB8), which were selective inhibitors
Bijo Mathew et al.
The journal of physical chemistry. B, 121(6), 1186-1203 (2017-01-14)
Selective monoamine oxidase-B (MAO-B) inhibitors are imperative in the treatment of various neurodegenerative disorders. Herein, we describe the pharmacophore generation and atom-based three-dimensional quantitative structure-activity relationship (3D-QSAR) analyses of previously reported thiophene-based hMAO-B inhibitors by our research group. The aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service