Skip to Content
Merck
All Photos(1)

Key Documents

SML2039

Sigma-Aldrich

SAK3

≥98% (HPLC)

Synonym(s):

8′-Methyl-2′,4-dioxo-2-(1-piperidinyl)-spiro[2-cyclopentene-1,3′(2′H)-imidazo[1,2-a]pyridine]-3-carboxylic acid ethyl ester, Ethyl 8′-methyl-2′,4-dioxo-2-(piperidin-1-yl)-2′H-spiro[cyclopent[2]ene-1,3′-imidazo[1,2-a]pyridine]-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N3O4
CAS Number:
Molecular Weight:
369.41
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OCC)C1=C(C2(CC1=O)N3C=CC=C(C3=NC2=O)C)N4CCCCC4

InChI key

WZHBYTVHUFURPY-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and orally active Cav3.1 & Cav3.3 T-type voltage-gated Ca2+ channel (T-VGCC) enhancer with in vivo cognitive deficits-improving efficacy.
SAK3 is a potent and orally active spiroimidazopyridine derivative that enhances Cav3.1 and Cav3.3, but not Cav3.2, T-type voltage-gated Ca2+ channel (T-VGCC) activity (∼20% peak current increase with 0.1 nM SAK3 by whole-cell patch-clamp using human Cav3.1 or Cav3.3 transfected neuro2A; no effect up to 10 nM using Cav3.2 transfectant). Acute SAK3 oral administration (0.5 mg/kg) significantly enhances hippocampal CA1 region acetylcholine (ACh) release in naïve mice, as well as increases hippocampal ACh levels and improves memory deficits among olfactory-bulbectomized (OBX) mice. Daily SAK3 oral administration 24 hrs following hypothyroidism induction by methimazole (MMI) is efficacious against MMI-induced medial septum (MS) cholinergic neurons loss (0.5-1 mg/kg/day) and cognitive deficits (1 mg/kg/day).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Husain Noreen et al.
Neurochemistry international, 108, 91-99 (2017-03-11)
Methimazole (MMI) is a first-line therapy used to manage hyperthyroidism and Graves' disease. Despite its therapeutic benefit, chronic MMI administration can lead to hypothyroidism and perturb brain homeostasis in patients, resulting in neuropsychiatric disorders such as depression and cognitive dysfunction.
Udaiyappan Janakiraman et al.
Neurobiology of disease, 143, 105006-105006 (2020-07-06)
T-type calcium channels, in the central nervous system, are involved in the pathogenesis of many neurodegenerative diseases, including TAF1 intellectual disability syndrome (TAF1 ID syndrome). Here, we evaluated the efficacy of a novel T-type Ca2+ channel enhancer, SAK3 (ethyl 8'-methyl-2'
Yasushi Yabuki et al.
Neuropharmacology, 117, 1-13 (2017-01-18)
T-type voltage-gated Ca
Chinnasamy Dhanalakshmi et al.
Neurobiology of disease, 149, 105224-105224 (2020-12-29)
The TATA-box binding protein associated factor 1 (TAF1) is part of the TFIID complex that plays a key role during the initiation of transcription. Variants of TAF1 are associated with neurodevelopmental disorders. Previously, we found that CRISPR/Cas9 based editing of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service