Skip to Content
Merck
All Photos(1)

Key Documents

SML0798

Sigma-Aldrich

Osanetant

≥98% (HPLC)

Synonym(s):

(R)-N-{{3-[1-Benzoyl-3-(3,4-dichlorophenyl)piperidin-3-yl]prop-1-yl}-4-phenylpiperidin-4-yl}-N-methylacetamine, SR-142801, SR142801

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H41Cl2N3O2
CAS Number:
Molecular Weight:
606.62
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +17 to +24°, c = 1 in methanol

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1c(ccc(c1)[C@@]4(CN(CCC4)C(=O)c5ccccc5)CCCN2CCC(CC2)(N(C)C(=O)C)c3ccccc3)Cl

InChI

1S/C35H41Cl2N3O2/c1-27(41)38(2)35(29-13-7-4-8-14-29)19-23-39(24-20-35)21-9-17-34(30-15-16-31(36)32(37)25-30)18-10-22-40(26-34)33(42)28-11-5-3-6-12-28/h3-8,11-16,25H,9-10,17-24,26H2,1-2H3/t34-/m0/s1

InChI key

DZOJBGLFWINFBF-UMSFTDKQSA-N

Biochem/physiol Actions

Osanetant is a selective antagonist of neurokinin 3 receptor NK3.
Osanetant is a selective antagonist of neurokinin 3 receptor NK3. Neurokinins (tachykinins) are members of a family of at least three neuropeptides, substance P, neurokinin A, and neurokinin B (NKB), with each mediating their biological effects through binding to a preferred G-protein-coupled receptor termed NK1, NK2, or NK3, respectively. All three NK receptors are expressed in regions of the central nervous system that are related to emotion and cognition (i.e., amygdala and hippocampus) and have been linked to various degrees in psychiatric disorders. Neurokinin receptors, including NK3 receptors, are also expressed in the motor and sensory systems of the digestive tract. Osanetant was in clinical trials for both irritable bowel syndrome and schizophrenia.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service