Skip to Content
Merck
All Photos(2)

Key Documents

SAE0198

Sigma-Aldrich

Calprotectin, human recombinant

Stabilized heterodimer of fusion tagged S100A9 and S100A8

Synonym(s):

MRP8/14 and S100A8/A9 complex

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

shipped in

wet ice

storage temp.

−20°C

General description

This recombinant human calprotectin is a heterodimer composed of tagged versions of both S100A8 and S100A9, where a 6xHis-SUMO tag is added to the N-terminus of S100A9, and a 6xHIS tag is added to the N-terminus of S100A8. This tagged version was found to have enhanced stability as compared to untagged versions of this protein. Removal of the tags can be achieved using HRV-3C protease (SAE0045 or SAE0110).

Application

Fecal Calprotectin is often used as a biomarker to identify inflammation in gastrointestinal disease1. This product is a recombinant version of the human protein found endogenously.

Biochem/physiol Actions

Calprotectin is a calcium and zinc binding protein that belongs to the S100 family. It is a heterodimer protein composed of S100A8 and S100A9.1 The protein is expressed in myeloid cells and in circulating monocytes and neutrophils, where it accounts for 60% of the cytosolic protein content. Calprotectin was found to play major roles in inflammation and the immune response by activation of NF-kB pathway via TLR4 and AGER,2 and its antimicrobial activity possibly through chelating of Zn.3 High concentrations of calprotectin are found and serve as a biomarker in rheumatoid, cystic fibrosis, inflammatory bowel disease and in Crohn′s disease.4 A meta-analysis also identified calprotectin as a potential biomarker for COVID-19.5

Physical form

The product is lyophilized from a sterile 0.2 µm filtered solution containing 12.5 mM Tris-Cl (pH 8.0), 150 mM NaCl, and 1 mM DTT.

Preparation Note

Reconstitute lyophilized vial in DMEM or in PBS to a concentration up to 500 µg/mL. Spin down to collect material. Reconstituted product should be stored in working aliquots at –20 °C. Do not store in a frost-free freezer. Avoid freeze-thaw cycles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service