Skip to Content
Merck
All Photos(1)

Key Documents

SAB4301741

Sigma-Aldrich

Anti-APLN

affinity isolated antibody

Synonym(s):

APEL, XNPEP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
IHC
species reactivity:
rat, human, mouse
technique(s):
immunohistochemistry: 1:20-1:100
citations:
3

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: 1:20-1:100

accession no.

NP_059109

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APLN(8862)

Immunogen

Synthetic peptide of human APLN

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayushmita Dutta et al.
Cell biology international (2024-04-18)
Apelin and its receptor (APJ) are expressed in the reproductive organs of some mammalian females. The function of oviduct has also been suggested to be compromised in the hyperandrogenism condition. However, expression of apelin and APJ has not been shown
Borgohain Anima et al.
Molecular reproduction and development, 90(1), 42-52 (2022-12-03)
Postnatal ovarian and uterine development is crucial to accomplished female fertility. Thus, the investigations of factors that present in pre-pubertal stages are important as it might be responsible for the regulation of ovarian and uterine function. Apelin, an adipokine and
Borgohain Anima et al.
Neuroendocrinology, 114(3), 234-249 (2023-10-30)
Apelin is an endogenous peptide, whose expression has been shown in the hypothalamus, pituitary, and ovary; furthermore, it is also called a neuropeptide, binding to apelin receptor (APJ) for various functions. It has been suggested that the hypothalamus, pituitary, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service