Skip to Content
Merck
All Photos(2)

Key Documents

S9924

Sigma-Aldrich

Anti-Syntaxin 4 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB CL
clone:
polyclonal
species reactivity:
rat
citations:
4
technique(s):
immunohistochemistry: suitable
western blot (chemiluminescent): 1:300-1:600

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunohistochemistry: suitable
western blot (chemiluminescent): 1:300-1:600

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STX4(6810)
mouse ... Stx4a(20909)
rat ... Stx4(81803)

Immunogen

synthetic peptide corresponding to amino acids 2-23 of rat or mouse syntaxin 4 (with additional C-terminal cysteine).

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Bennett et al.
Cell, 74(5), 863-873 (1993-09-10)
Syntaxins A and B are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. A family of syntaxin-related proteins from rat has been identified that shares 23%-84% amino acid identity. Each of the six
J T Tellam et al.
The Journal of biological chemistry, 272(10), 6179-6186 (1997-03-07)
We have previously identified three mammalian Sec1/Munc-18 homologues in adipocytes (Tellam, J. T., McIntosh, S., and James, D. E. (1995) J. Biol. Chem. 270, 5857-5863). These proteins are thought to modulate the interaction between vesicle membrane and target membrane soluble
Kyubo Kim et al.
The Biochemical journal, 446(3), 383-394 (2012-06-15)
Airway mucin secretion and MC (mast cell) degranulation must be tightly controlled for homoeostasis of the lungs and immune system respectively. We found the exocytic protein Munc18b to be highly expressed in mouse airway epithelial cells and MCs, and localized
Shifeng Wang et al.
Traffic (Copenhagen, Denmark), 21(10), 636-646 (2020-08-28)
Exocytosis is a vesicle fusion process driven by soluble N-ethylmaleimide-sensitive factor attachment protein receptors (SNAREs). A classic exocytic pathway is insulin-stimulated translocation of the glucose transporter type 4 (GLUT4) from intracellular vesicles to the plasma membrane in adipocytes and skeletal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service