Skip to Content
Merck
All Photos(1)

Key Documents

RAB0623

Sigma-Aldrich

Human BACE1 / Beta-secretase 1 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 ng/mL
standard curve range: 1.024-250 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BACE1(23621)

General description

The BACE-1 ELISA kit provides for the Quantitative measurement of BACE-1 in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunyan Ye et al.
Frontiers in aging neuroscience, 12, 555594-555594 (2020-11-17)
Background: The mechanism underlying delirium, a common acute fluctuating mental state, may be related to the activation of a neuroinflammatory response. In this study, we attempted to investigate whether plasma inflammatory response markers, vascular and cerebrovascular injury-related markers, and neurodegeneration-associated
Bo Hong et al.
Experimental and therapeutic medicine, 16(4), 3694-3702 (2018-09-21)
Gestational hypertensive disorder is a complication of pregnancy, which adversely affects mother-child health. Captopril and prazosin are two agents that are frequently applied for the treatment of patients with gestational hypertension. However, the cooperative efficacy of captopril and prazosin has
Xiaoyao Zheng et al.
Acta biomaterialia, 49, 388-401 (2016-11-16)
To realize the therapeutic potential of gene drugs for Alzheimer's disease (AD), non-invasive, tissue-specific and efficient delivery technologies must be developed. Here, a hybrid system for amyloid plaques targeted siRNA delivery was formed by PEGylated Poly(2-(N,N-dimethylamino) ethyl methacrylate) (PEG-PDMAEMA) conjugated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service