Skip to Content
Merck
All Photos(2)

Key Documents

RAB0213

Sigma-Aldrich

Human HGF ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type tissue lysate
sample type cell lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HGF(3082)

General description

The Human HGF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human HGF cell lysate and tissue lysate.

Immunogen

Recombinant Human HGF

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Wen et al.
Journal of translational medicine, 12, 114-114 (2014-06-03)
Autologous transplantation of modified mesenchymal stem cells (MSCs) is a promising candidate for the treatment of the refractory clinical disease, avascular necrosis of the femoral head (ANFH). Our previous attempts by compounding MSCs with medical fibrin glue to treat ANFH
Jesse Hudson et al.
Molecular and cellular biology, 34(19), 3689-3701 (2014-07-30)
Breast cancers are stratified into distinct subtypes, which influence therapeutic responsiveness and patient outcome. Patients with luminal breast cancers are often associated with a better prognosis relative to that with other subtypes. However, subsets of patients with luminal disease remain
Elisa Tinazzi et al.
Immunology letters, 161(2), 236-240 (2014-01-23)
CD30 and CD30 ligand (CD30L) are members of TNF-receptor and TNF superfamilies respectively. CD30(+)T cells are increased in several diseases and interaction between CD30(+) and CD30L(+)T cells leads either to cell proliferation or apoptosis. In patients with rheumatoid arthritis (RA)
Ji-Dong Chen et al.
Asian Pacific journal of cancer prevention : APJCP, 15(11), 4545-4548 (2014-06-28)
To investigate the changes of serum vascular endothelial growth factor (VEGF), soluble interleukin-2 receptor (SIL-2R) and hepatocyte growth factor (HGF) contents in patients with primary hepatocellular carcinoma (HCC) before and after percutaneous microwave coagulation therapy (PMCT) and determine their clinical
Mishaal B Patel et al.
Carcinogenesis, 35(8), 1891-1900 (2014-05-31)
Activated cancer-associated human pancreatic stellate cells (CAhPSCs, which produce the collagenous stroma of pancreatic cancer [PC]) are known to play a major role in PC progression. Apart from inducing cancer cell proliferation and migration, CAhPSCs have also been implicated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service