RAB0024
Human β-2 Microglobulin ELISA Kit
for serum, plasma, cell culture supernatant and urine
Synonym(s):
B2M
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.84
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type serum
sample type urine
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 15.6-1000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... B2M(567)
General description
β2-microglobulin (B2M) is a non-glycosylated polypeptide composed of 99 amino acids. It is present on the surface of all nucleated cells. The protein is characterized with a single polypeptide chain, which is linked noncovalently to major histocompatibility complex (MHC) class I cell surface antigen. The gene coding for B2M is mapped to human chromosome 15q.
The Human BDNF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human BDNF in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human β-2-Microglobulin
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
β2-microglobulin (B2M) plays a vital role in cell survival, proliferation and metastasis in various types of cancer. Elevated concentration of β-2-microglobulin (B2M) can be considered as a prognostic marker for hemophagocytic lymphohistiocytosis (HLH). Elevated concentration of circulating B2M, due to renal dysfunction and long term hemodialysis, leads to dialysis-related amyloidosis. Overexpression of serum B2M is observed in hepatitis C virus (HCV) related chronic liver diseases, therefore it might act as a potential marker for the HCV disease progression towards cirrhosis and carcinoma. Missense mutation in the B2M gene results in familial hypercatabolic hypoproteinemia (FHH).
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lengths of chromosomal segments conserved since divergence of man and mouse.
Nadeau JH and Taylor BA.
Proceedings of the National Academy of Sciences of the USA, 81(3), 814-818 (1984)
The Prognostic Significance of Beta2 Microglobulin in Patients with Hemophagocytic Lymphohistiocytosis.
Jiang T
Disease Markers, 1523959 (2016)
Serum Beta-2 Microglobulin: a Possible Marker for Disease Progression in Egyptian Patients with Chronic HCV Related Liver Diseases.
Ouda SM
Asian Pacific Journal of Cancer Prevention, 16(17), 7825-7829 (2015)
Kinetics of FcRn-mediated recycling of IgG and albumin in human: pathophysiology and therapeutic implications using a simplified mechanism-based model.
Kim J
Clinical Immunology (Orlando, Fla.), 122(2), 146-155 (2007)
Uncovering the Early Assembly Mechanism for Amyloidogenic ?2-Microglobulin Using Cross-linking and Native Mass Spectrometry.
Hall Z
The Journal of Biological Chemistry, 291(9), 4626-4637 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service