PZ0102
CP-93129 dihydrochloride hydrate
≥98% (HPLC)
Synonym(s):
1,4-Dihydro-3-(1,2,3,6-tetrahydro-4-pyridinyl)-5H-pyrrol[3,2-b]pyridin-5-one dihydrochloride hydrate
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
gray
solubility
deionized water: ≥5 mg/mL
storage temp.
2-8°C
SMILES string
O.Cl.Cl.O=C1Nc2c(C=C1)[nH]cc2C3=CCNCC3
InChI
1S/C12H13N3O.2ClH.H2O/c16-11-2-1-10-12(15-11)9(7-14-10)8-3-5-13-6-4-8;;;/h1-3,7,13-14H,4-6H2,(H,15,16);2*1H;1H2
InChI key
UXDAADMPPAHNMY-UHFFFAOYSA-N
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
We offer many products related to serotonin receptors for your research needs.
We offer many products related to serotonin receptors for your research needs.
We offer many products related to serotonin receptors for your research needs.
We offer many products related to serotonin receptors for your research needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service