Skip to Content
Merck
All Photos(2)

Key Documents

PRS3627

Sigma-Aldrich

Anti-LAMP-2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Lysosomal-associated membrane protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Lamp2(16784)

General description

Lamp2 (lysosome-associated membrane protein gene)is located in the lysosomal lumen and lysosomal membrane. However upon chronic acidosis in the tumour microenvironment, it is found to be present in the plasma membrane.

Immunogen

a 17 amino acid peptide from near the center of human LAMP-2.

Application

Anti-LAMP-2 antibody has been used in immunofluorescence and western blot.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

LAMP2 (lysosome-associated membrane protein gene) preserves lysosomal membranes from acid proteolysis. It serves as an acidity biomarker in spheroids and xenografts. It also has some role in maturation of autophagic vacuoles and cell-cell or cell-extracellular matrix adhesion. Reduction of LAMP2 (lysosome-associated membrane protein gene) can increase acidosis-mediated toxicity. Mutation in LAMP2 can cause cardiomyopathy in young patients.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3627.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of white light-emitting diode (LED) exposure on retinal pigment epithelium in vivo
Jaadane I, et al.
Journal of Cellular and Molecular Medicine (2017)
Unique properties of lamp2a compared to other lamp2 isoforms
Cuervo AM and Dice JF
Journal of Cell Science, 113 Pt 24, 4441-4450 (2000)
Chronic acidosis in the tumour microenvironment selects for overexpression of LAMP2 in the plasma membrane
Damaghi M, et al.
Nature Communications, 6:8752 (2015)
Imene Jaadane et al.
Journal of cellular and molecular medicine, 21(12), 3453-3466 (2017-07-01)
Ageing and alteration of the functions of the retinal pigment epithelium (RPE) are at the origin of lost of vision seen in age-related macular degeneration (AMD). The RPE is known to be vulnerable to high-energy blue light. The white light-emitting
Characterization of stress response in human retinal epithelial cells.
Giansanti V
Journal of Cellular and Molecular Medicine, 17(1), 103-115 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service