Skip to Content
Merck
All Photos(1)

Key Documents

P9180

Sigma-Aldrich

Procaterol hydrochloride

Synonym(s):

erythro-8-Hydroxy-5-[1-hydroxy-2-(isopropylamino)butyl]-2(1H)-quinolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N2O3 · HCl
CAS Number:
Molecular Weight:
326.82
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

powder

storage temp.

2-8°C

SMILES string

Cl.CC[C@H](NC(C)C)[C@H](O)c1ccc(O)c2NC(=O)C=Cc12

InChI

1S/C16H22N2O3.ClH/c1-4-12(17-9(2)3)16(21)11-5-7-13(19)15-10(11)6-8-14(20)18-15;/h5-9,12,16-17,19,21H,4H2,1-3H3,(H,18,20);1H/t12-,16+;/m0./s1

InChI key

AEQDBKHAAWUCMT-CVHDTDHSSA-N

General description

Procaterol is used to prevent lung fibroblast migration. It has anti-inflammatory properties. It can block type 2 T helper (Th2)-related chemokine production in human monocytes and bronchial epithelial cells.

Biochem/physiol Actions

β2-adrenoceptor agonist; bronchodilator.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukiko Hiraguchi et al.
International archives of allergy and immunology, 155 Suppl 1, 85-89 (2011-06-10)
Viral respiratory tract infections play an important role in the inception and exacerbation of asthma. Eosinophils, major effector cells in asthma, often accumulate in the airways during viral infections and are possibly activated by respiratory RNA viruses through Toll-like receptor
Masahiro Satake et al.
Arzneimittel-Forschung, 61(1), 8-13 (2011-03-02)
The purpose of this study was to evaluate the inhibitory effect of procaterol (procaterol hydrochloride, CAS 62929-91-3) on exercise dynamic lung hyperinflation during the 6-min walk test (6MWT) in stable chronic obstructive disease (COPD) patients. Fourteen patients with stable COPD
Ethanolamines: Advances in Research and Application (2012)
F Preitner et al.
British journal of pharmacology, 124(8), 1684-1688 (1998-10-02)
The beta3-adrenoceptor plays an important role in the adrenergic response of brown and white adipose tissues (BAT and WAT). In this study, in vitro metabolic responses to beta-adrenoceptor stimulation were compared in adipose tissues of beta3-adrenoceptor knockout and wild type
Y Yamazaki et al.
British journal of pharmacology, 124(3), 593-599 (1998-07-01)
1. The beta-adrenoceptor (beta-AR) subtypes mediating relaxation of the rabbit, rat and canine detrusors were subjected to functional investigation using selective beta-AR agonists and antagonists. 2. In all three species, isoprenaline, noradrenaline and adrenaline each produced a concentration-dependent relaxation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service