Skip to Content
Merck
All Photos(1)

Key Documents

P9120

Sigma-Aldrich

PNGase F from Elizabethkingia meningoseptica

recombinant, expressed in E. coli, set of 100 units nanomolar unit

Synonym(s):

N-Glycanase®, N-Glycosidase F, PNGase F from Chryseobacterium meningosepticum, PNGase F from Flavobacterium meningosepticum, Peptide N-glycosidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

conjugate

(N-linked)

specific activity

≥10 units/mg protein

mol wt

36 kDa

packaging

set of 100 units nanomolar unit

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

PNGase F from Elizabethkingia meningoseptica has been used in deglycosylation
  • of recombinant soybean agglutinin (rSBA) in Nicotiana benthamiana (NbrSBA) and Solanum tuberosum (StrSBA)[1]
  • of frontal cortical lysate to verify the glycosylation profile of β-secretase (BACE proteins)[2]
  • of cell lysate for evaluating the siRNA silencing of cellular prion protein (PrPc) post transfection[3]

PNGase F is a glycosylasparaginase used to deglycosylate proteins. It is widely used in structure-function studies of glycoproteins[4].
Used to deglycosylate protein.

Biochem/physiol Actions

Cleaves an entire glycan from a glycoprotein provided the glycosylated asparagine moiety is substituted on its amino and carboxyl terminus with a polypeptide chain.
PNGase F cleaves an entire glycan from a glycoprotein provided the glycosylated asparagine moiety is substituted on its amino and carboxyl terminus with a polypeptide chain. It deaminates the asparagine to aspartic acid, but leaves the oligosaccharide intact. PNGase F will not remove oligosaccharides containing α(1-3)-linked core fucose, commonly found in plant glycoproteins. A tripeptide with the oligosaccharide-linked asparagine as the central residue is the minimal substrate for PNGase F[4].

Packaging

Each set includes enzyme, two formulations of 5× reaction buffer (for routine and mass spectrometry downstream analysis), detergent and denaturation solutions

Unit Definition

One unit will catalyze the release of N-linked oligosaccharides from 1 micromole of denatured ribonuclease B in one minute at 37°C at pH 7.5 monitored by SDS-PAGE. One Sigma unit of PNGase F activity is equal to 1 IUB milliunit.

Legal Information

N-Glycanase is a registered trademark of Agilent Technologies Inc

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PrPc activation induces neurite outgrowth and differentiation in PC12 cells: role for caveolin-1 in the signal transduction pathway
Pantera B, et al.
Journal of Neurochemistry, 110(1), 194-207 (2009)
Mitochondrial respiratory inhibition and oxidative stress elevate beta-secretase (BACE1) proteins and activity in vivo in the rat retina
Xiong K, et al.
Experimental Brain Research. Experimentelle Hirnforschung. Experimentation Cerebrale, 181(3), 435-446 (2007)
G E Norris et al.
Structure (London, England : 1993), 2(11), 1049-1059 (1994-11-15)
Peptide:N-glycosidase F (PNGase F) is an enzyme that catalyzes the complete removal of N-linked oligosaccharide chains from glycoproteins. Often called an endoglycosidase, it is more correctly termed an amidase or glycosylasparaginase as cleavage is at the asparagine-sugar amide linkage. The
Reynald Tremblay et al.
Transgenic research, 20(2), 345-356 (2010-06-19)
Soybean agglutinin (SBA) is a specific N-acetylgalactosamine-binding plant lectin that can agglutinate a wide variety of cells. SBA has great potential for medical and biotechnology-focused applications, including screening and treatment of breast cancer, isolation of fetal cells from maternal blood
Anna C Need et al.
Journal of medical genetics, 49(6), 353-361 (2012-05-15)
There is considerable interest in the use of next-generation sequencing to help diagnose unidentified genetic conditions, but it is difficult to predict the success rate in a clinical setting that includes patients with a broad range of phenotypic presentations. The

Articles

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service