Skip to Content
Merck
All Photos(2)

Key Documents

P5436

Sigma-Aldrich

Sodium propionate

≥99.0%, BioReagent, suitable for insect cell culture

Synonym(s):

Propionic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COONa
CAS Number:
Molecular Weight:
96.06
Beilstein:
3566934
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.75
biological source:
synthetic (organic)

biological source

synthetic (organic)

Quality Level

product line

BioReagent

Assay

≥99.0%

form

powder

technique(s)

cell culture | insect: suitable

mp

285-286 °C (lit.)

solubility

H2O: 1 g/mL, clear, colorless to very faintly yellow

SMILES string

[Na+].CCC([O-])=O

InChI

1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1

InChI key

JXKPEJDQGNYQSM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used as a mold inhibitor in insect cell culture applications.

Biochem/physiol Actions

Sodium propionate has an ability to prevent ketosis in dairy cattle.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of sodium propionate in the prevention of ketosis in dairy cattle
Schultz LH
Journal of Dairy Science, 41(1), 160-168 (1958)
Zhenhua Zhou et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 41(2), 267-281 (2020-03-11)
Sodium butyrate, a short-chain fatty acid, is predominantly produced by gut microbiota fermentation of dietary fiber and serves as an important neuromodulator in the central nervous system. Recent experimental evidence has suggested that sodium butyrate may be an endogenous ligand
Elena Zagato et al.
Nature microbiology, 5(3), 511-524 (2020-01-29)
The microbiota has been shown to promote intestinal tumourigenesis, but a possible anti-tumourigenic effect has also been postulated. Here, we demonstrate that changes in the microbiota and mucus composition are concomitant with tumourigenesis. We identified two anti-tumourigenic strains of the
Benjamin Pontiller et al.
Frontiers in microbiology, 11, 588778-588778 (2020-10-27)
Bacteria play a key role in the planetary carbon cycle partly because they rapidly assimilate labile dissolved organic matter (DOM) in the ocean. However, knowledge of the molecular mechanisms at work when bacterioplankton metabolize distinct components of the DOM pool
Bistra B Nankova et al.
PloS one, 9(8), e103740-e103740 (2014-08-30)
Alterations in gut microbiome composition have an emerging role in health and disease including brain function and behavior. Short chain fatty acids (SCFA) like propionic (PPA), and butyric acid (BA), which are present in diet and are fermentation products of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service