Key Documents
P0924
Microcapillary tube, calibrated
capacity 40 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
glass
packaging
pack of 250 ea
manufacturer/tradename
Drummond Scientific 2-000-040
measuring range
accuracy: ±0.5%
capacity
40 μL
color
color-coded yellow
General description
  • Drummond Calibrated Micropipettes provide accuracy and convenience for a wide variety of liquid transfer applications.
  • Made of high quality glass.
  • Fire polished micropipettes are imprinted with volume and a color-code bar for easy identification.
  • Calibrated "To Deliver."
High quality Drummond micropipettes are fire polished, imprinted with volume and a color-code bar for easy identification. Calibrated "To Deliver."
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What is the diameter of pipette? I need approxemetly tip diameter 300 μm
    1 answer
    1. This product has an OD of 1.372 mm and an ID of 936 um. The smallest option for the Drummond calibrated microcapillary tubes is product P0549, a 1 - 5 uL volume tube. This item has an OD of 1.092 mm and an ID of 340 um. Please see the link below to review this product option:
      https://www.sigmaaldrich.com/product/sigma/p0549
      Helpful?