MSST0054
SILu™Lite AKT3, RAC-gamma serine/threonine-protein kinase human
recombinant, expressed in HEK 293 cells, MS Protein Standard
Synonym(s):
Protein kinase Akt-3, Protein kinase B gamma (PKB gamma), RAC-PK-gamma, STK-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23201100
NACRES:
NA.12
Recommended Products
recombinant
expressed in HEK 293 cells
Quality Level
tag
His tagged
Assay
≥95% (SDS-PAGE)
form
liquid
suitability
suitable for mass spectrometry (internal calibrator)
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... AKT3(10000)
Related Categories
General description
SILu™Lite AKT3 is a recombinant human protein expressed in human 293 cells. It consists of 500 amino acids (including N-terminal polyhistidine and flag tags), with a calculated molecular mass of 58.2 kDa. SILu™Lite AKT3 is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.
Sequence
MDYKDDDDKGHHHHHHHHGGQMSDVTIVKEGWVQKRGEYIKNWRPRYFLLKTDGSFIGYKEKPQDVDLPYPLNNFSVAKCQLMKTERPKPNTFIIRCLQWTTVIERTFHVDTPEEREEWTEAIQAVADRLQRQEEERMNCSPTSQIDNIGEEEMDASTTHHKRKTMNDFDYLKLLGKGTFGKVILVREKASGKYYAMKILKKEVIIAKDEVAHTLTESRVLKNTRHPFLTSLKYSFQTKDRLCFVMEYVNGGELFFHLSRERVFSEDRTRFYGAEIVSALDYLHSGKIVYRDLKLENLMLDKDGHIKITDFGLCKEGITDAATMKTFCGTPEYLAPEVLEDNDYGRAVDWWGLGVVMYEMMCGRLPFYNQDHEKLFELILMEDIKFPRTLSSDAKSLLSGLLIKDPNKRLGGGPDDAKEIMRHSFFSGVNWQDVYDKKLVPPFKPQVTSETDTRYFDEEFTAQTITITPPEKYDEDGMDCMDNERRPHFPQFSYSASGRE
Physical form
Supplied as a liquid containing phosphate buffered saline.
Legal Information
SILu is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service