Skip to Content
Merck
All Photos(5)

Key Documents

M8434

Sigma-Aldrich

Anti-Mcl-1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40-42 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): 100-150 μg using lysate of mitochondria from 2.5 to 5.0 × 107 HeLa cells
microarray: suitable
western blot: 1:8,000 using a HeLa cell (human epithelioid carcinoma) mitochondria extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Anti-Mcl-1 is developed in rabbit using a synthetic peptide corresponding to an internal region of Mcl-1 of human origin with N-terminal added cysteine, conjugated to maleimide activated keyhole limpet hemocyanin (KLH), as immunogen. MCL1 apoptosis regulator, BCL2 family member (Mcl-1) is expressed in many normal and neoplastic cells and is especially abundant in skeletal and cardiac muscle and in germinal centers of lymphoid tissues.
Mcl-1 is a member of Bcl-2 family that contains 3 Bcl-2 homology (BH) domains, BH1, BH2 and BH3, originally identified as an upregulated gene in human myeloid leukemia cell line (ML-1) in response to PMA. It is regulated transcriptionally and post-transcriptionally to enhance the cell survival. [1] Mcl-1 is induced rapidly through cytokine-mediated survival pathways. Additionally, the upstream half of Mcl-1 RNA contains PEST (pro (P) Glu (E), Ser (S) and Thr (T) ) sequences and exhibits rapid turn-over. Increased expression of Mcl-1 maintains cell viability, decreased expression promotes cell death. Mcl-1 is reported to exhibit differentiation stage-specific expression in hematopoietic lineages and epithelial cells. A splice variant of Mcl-1, Mcl-1s promotes cell death. [1] Often, the expression of Mcl-1 is induced via the MAPK signalling pathway acting on SRF/Elk-1 or Akt/CREB regulated pathway. Mcl-1 is expressed in many normal and neoplastic cells and is especially abundant in skeletal and cardiac muscle and in germinal centers of lymphoid tissues. It is predominantly expressed in the mitochondria but in neutrophils it seems to be mainly located in nuclear fractions. [1]

Specificity

Anti-Mcl-1 specifically recognizes Mcl-1 in tissue and cell extracts (40 to 42 kDa doublet).

Immunogen

synthetic peptide corresponding to an internal region of Mcl-1 of human origin (amino acids 121-139). This sequence is highly similar in mouse and rat.

Application

A minimum working dilution of 1:8000 is determined by immunoblotting using HeLa human epithelioid carcinoma mitochondria extract. It may also be used for detection by immunoblotting in Human melanoma cell lines, Human colon adenocarcinoma HT-29 and SW620 cells. Mcl-1 is immunoprecipitated from the lysate of mitochondria from 2.5 to 5.0x10 7 HeLa cells using 100 to 150 μg of the antibody. The antibody is suitable for protein microarray applications.
Anti-Mcl-1 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

MCL1 apoptosis regulator, BCL2 family member (Mcl-1) exhibits great lability presumably due to its PEST sequence (P, Pro; E, Glu; S, Ser; T, Thr). Unlike the stable Bcl-2 protein, Mcl-1 exhibits great lability presumably due to its PEST sequence (P, Pro; E, Glu; S, Ser; T, Thr). Mcl-1, like Bcl-2, promotes cell viability under conditions which otherwise cause apoptosis. MCL1 acts as a chaperone of fortilin by binding and stabilizing it. It also interacts and negatively regulates the proliferating cell nuclear antigen (PCNA).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kung-Wen Lu et al.
International journal of molecular sciences, 23(18) (2022-09-24)
Some clinically used anti-cancer drugs are obtained from natural products. Allyl isothiocyanate (AITC), a plant-derived compound abundant in cruciferous vegetables, has been shown to possess an anti-cancer ability in human cancer cell lines in vitro, including human brain glioma cells.
D A Moulding et al.
Blood, 92(7), 2495-2502 (1998-09-25)
Human neutrophils possess a very short half-life because they constitutively undergo apoptosis. Cytokines, such as granulocyte-macrophage colony-stimulating factor (GM-CSF), and other agents can rescue neutrophils from apoptosis but the molecular mechanisms involved in this rescue are undefined. Here, we show
Differences in TRAIL-induced changes of Mcl-1 expression among distinct human colon epithelial cell lines
Vaculova A, et al.
Experimental Cell Research, 315(19), 3259-3266 (2009)
Exon skipping in Mcl-1 results in a bcl-2 homology domain 3 only gene product that promotes cell death
Bingle CD, et al.
Test, 275(29), 22136-22146 (2000)
S Krajewski et al.
The American journal of pathology, 146(6), 1309-1319 (1995-06-01)
The mcl-1 gene encodes an approximately 37-kd protein that has significant homology with Bcl-2, an inhibitor of programmed cell death that is expressed in many types of long-lived cells. In this study we determined the in vivo patterns of Mcl-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service