Skip to Content
Merck
All Photos(2)

Key Documents

M5696

Sigma-Aldrich

Myoglobin from equine skeletal muscle

BioUltra, 95-100% (SDS-PAGE)

Synonym(s):

Myoglobin from horse skeletal muscle

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

equine skeletal muscle

product line

BioUltra

Assay

95-100% (SDS-PAGE)

form

essentially salt-free, lyophilized powder

mol wt

~17 kDa(lit.)

purified by

affinity chromatography

Iron content content

~0.30%

technique(s)

mass spectrometry (MS): suitable

solubility

H2O: soluble 10 mg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

horse ... MB(100054434)

Looking for similar products? Visit Product Comparison Guide

Application

Myoglobin from equine skeletal muscle has been used to induce acute kidney injury (AKI) in mice. It has also been used as a standard for top down mass spectrometry.
Myoglobin is used as a molecular weight marker and a standard for mass spectroscopy and X-ray crystallography.Myoglobin from equine skeletal muscle was used in a study to test experimental protein mixture for validating tandem mass spectral analysis.

Biochem/physiol Actions

Myoglobin from horse skeletal muscle is a single chain heme protein containing 153 amino acid residues. It possesses no disulfide bridges or free -SH groups. Myoglobin contains 8 variously sized right-handed helical regions, joined by non-ordered or random coil regions.
Myoglobin is critical to skeletal muscle O2 supply at near-maximum oxygen demand, and prevents anoxia by maintaining PO2 above levels needed to support mitochondrial function.
Myoglobin is critical to skeletal muscle O2 supply at near-maximum oxygen demand, and prevents anoxia by maintaining PO2 above levels needed to support mitochondrial function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Powell et al.
Journal of chromatography, 317, 87-92 (1984-12-28)
Urine and serum myoglobin have been separated on an anion-exchange column, packed by the slurry technique. Urine or serum was injected directly into the column and eluted isocratically with Tris buffer. Freshly prepared myoglobin from human muscle gives two peaks
Seok Jong Song et al.
International journal of molecular sciences, 21(22) (2020-11-19)
A recent study showed that early renal tubular injury is ameliorated in Nod-like receptor pyrin domain-containing protein 3 (NLRP3) KO mice with rhabdomyolysis-induced acute kidney injury (RIAKI). However, the precise mechanism has not been determined. Therefore, we investigated the role
Z K Shihabi
Journal of chromatography. B, Biomedical applications, 669(1), 53-58 (1995-07-07)
Capillary electrophoresis was used in this study to separate urinary myoglobin from hemoglobin based on its electrophoretic mobility. Urine was applied directly without any treatment. The separation was accomplished in less than 7 min. Myoglobin extracted from human muscle tissues
Protein Bioinformatics (2017)
J Zaia et al.
Rapid communications in mass spectrometry : RCM, 6(1), 32-36 (1992-01-01)
Myoglobins from horse heart muscle, horse skeletal muscle and sperm whale are widely used as calibration standards or test compounds for various mass spectrometric methodologies. In all such cases reported in the literature, a molecular weight value is used (16,950.5

Articles

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

Professors summarize recent 2D materials synthesis advancements and biosensing applications in various fields.

Professors summarize recent 2D materials synthesis advancements and biosensing applications in various fields.

Professors summarize recent 2D materials synthesis advancements and biosensing applications in various fields.

See All

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service