M1901
Liquid scintillation vials, glass
volume 6 mL, Minivial, vial size 17 mm × 58 mm , with cap, Cap: 15mm urea with metal foil liner., pkg of 1000 ea
Synonym(s):
Glass Scintillation Vials
All Photos(1)
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
Recommended Products
material
glass vial
metal foil liner
feature
closure type white urea cap
packaging
pkg of 1000 ea
vial size
17 mm × 58 mm , with cap
volume
6 mL
fitting
thread for 15-425
General description
Wheaton 180 brand low-potassium glass yielding consistent low backgrounds and high UV transmission. White caps fit tightly and can be written on.
Packaging
Vials are shrink-wrapped, 200 per tray.
Caps are packed in a separate tray.
Caps are packed in a separate tray.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)