Skip to Content
Merck
All Photos(1)

Key Documents

M045000

Sigma-Aldrich

6-Tetrachloro-Fluorescein Phosphoramidite

Synonym(s):

6-(4, 7, 2′, 7′-Tetrachloro-3′,6′-dipivaloylfluoresceinyl-6-carboxamido)-hexyl-1-O-(2-cyanoethyl)-(N,N-diisopropyl)-phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H54N3O10Cl4P
Molecular Weight:
981.72
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

Assay

≥97% (31P-NMR)
≥97.0% (reversed phase HPLC)

Quality Level

color

white to off-white, powder

suitability

conforms to structure for H-NMR
conforms to structure for H-NMR

storage temp.

−70°C

SMILES string

O=C(C1=C(Cl)C=C(C(NCCCCCCOP(N(C(C)C)C(C)C)OCCC#N)=O)C(Cl)=C12)OC32C4=C(C=C(OC(C(C)(C)C)=O)C(Cl)=C4)OC5=CC(OC(C(C)(C)C)=O)=C(Cl)C=C53

InChI

1S/C46H54Cl4N3O10P/c1-25(2)53(26(3)4)64(59-19-15-16-51)58-18-14-12-11-13-17-52-40(54)27-20-32(49)37-38(39(27)50)46(63-41(37)55)28-21-30(47)35(61-42(56)44(5,6)7)23-33(28)60-34-24-36(31(48)22-29(34)46)62-43(57)45(8,9)10/h20-26H,11-15,17-19H2,1-10H3,(H,52,54)

InChI key

RKVIHIDUTMZQEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service