Skip to Content
Merck
All Photos(4)

Key Documents

L2158

Sigma-Aldrich

L-Leucine p-nitroanilide hydrochloride

chromogenic, ≥98% (HPLC), powder

Synonym(s):

H-Leu-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N3O3 · HCl
CAS Number:
Molecular Weight:
287.74
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.28

Product Name

L-Leucine p-nitroanilide hydrochloride, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

solubility

water: 25 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CC(C)C[C@H](N)C(=O)Nc1ccc(cc1)N(=O)=O

InChI

1S/C12H17N3O3.ClH/c1-8(2)7-11(13)12(16)14-9-3-5-10(6-4-9)15(17)18;/h3-6,8,11H,7,13H2,1-2H3,(H,14,16);1H/t11-;/m0./s1

InChI key

PFOYXOWPGRWPLS-MERQFXBCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Leucine p-nitroanilide hydrochloride has been used as a substrate to measure the activity of leucine aminopeptidase (LAP):
  • for spectrophotometric assays
  • in the intestinal samples, such as duodenum, jejunum, and ileum
  • in the hemolymph of Manila clam

Substrates

Substrate for the colorimetric determination of leucine aminopeptidase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan E Ford et al.
Fish & shellfish immunology, 23(2), 280-291 (2006-12-19)
Components of the haemolymph are understood to constitute the internal defense system of bivalve mollusks and their levels are often considered to be indicators of "health"; however, relatively little proof exists of the role that these elements play in the
J Williams et al.
British poultry science, 49(3), 329-339 (2008-06-24)
1. The objective of this experiment was to study two feeding methods, which could potentially act on the gut microflora, the structure and/or the function of the digestive tract and thereby improve the performance of broilers. 2. Four dietary treatments
Arun Kumar Singh et al.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases, 16(5), e375-e381 (2012-03-14)
Staphylococcus aureus has emerged as a major drug-resistant pathogen in hospital- and community-acquired infections. Leucine aminopeptidase (LAP) is known to be essential for survival of the bacteria; however the LAP of S. aureus has not been extensively characterized. In this
Characterization of proteolytic enzyme activities in macroalgae
Perez-Llorens JL, et al.
European Journal of Phycology, 38(1), 55-64 (2003)
Arun Kumar Singh et al.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases, 16(5), e375-e381 (2012-03-14)
Staphylococcus aureus has emerged as a major drug-resistant pathogen in hospital- and community-acquired infections. Leucine aminopeptidase (LAP) is known to be essential for survival of the bacteria; however the LAP of S. aureus has not been extensively characterized. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service