Skip to Content
Merck
All Photos(1)

Key Documents

JN0004

Sigma-Aldrich

Tolmetin sodium dihydrate

≥98% (HPLC)

Synonym(s):

Tolmetin sodium salt dihydrate, 1-Methyl-5-(4-methylbenzoyl)-1H-pyrrole-2-acetic acid sodium salt dihydrate, Sodium 2-(1-methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl)acetate dihydrate, 1-Methyl-5-(p-toluoyl)pyrrole-2-acetic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H14NO3Na · 2H2O
CAS Number:
Molecular Weight:
315.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

room temp

SMILES string

[Na+].[H]O[H].[H]O[H].Cc1ccc(cc1)C(=O)c2ccc(CC([O-])=O)n2C

InChI

1S/C15H15NO3.Na.2H2O/c1-10-3-5-11(6-4-10)15(19)13-8-7-12(16(13)2)9-14(17)18;;;/h3-8H,9H2,1-2H3,(H,17,18);;2*1H2/q;+1;;/p-1

InChI key

QQILXENAYPUNEA-UHFFFAOYSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Tolmetin sodium is a nonsteroidal anti-inflammatory drug (NSAID) that inhibits prostaglandin synthetase in vitro and reduces plasma levels of prostaglandin E. Tolmetin sodium exhibits anti-inflammatory, analgesic and antipyretic activity Its precise mechanism of is unknown. Tolmetin is used to treat rheumatoid arthritis, osteoarthrosis, pain, and ankylosing spondylitis.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Davies et al.
Current medical research and opinion, 7(2), 115-120 (1980-01-01)
A double-blind crossover study was carried out in 21 patients with osteoarthrosis of the hip to compare the effectiveness and tolerance of 1200 mg tolmetin sodium daily with 75 mg indomethacin daily. Each drug was given in 3 equally divided
F J Persico et al.
The Journal of pharmacology and experimental therapeutics, 247(3), 889-896 (1988-12-01)
The glycine amide of tolmetin sodium (TGA) functions as a prodrug and was demonstrated to be more potent than the parent compound as an inhibitor of developing and established adjuvant arthritis in the female Lewis rat. In contrast, the glycine
Afaf A Ramadan et al.
Journal of pharmaceutical investigation, 48(6), 673-683 (2019-01-01)
The objective of the present study was to develop rectal mucoadhesive hydrogels loaded with Tolmetin Sodium, a non-steroidal anti-inflammatory drug, for prolonged duration of action and increased bioavailability. Fourteen formulae were prepared with different types and concentrations of polymers as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service