Skip to Content
Merck
All Photos(1)

Key Documents

I6527

Sigma-Aldrich

Anti-Interleukin-4 Soluble Receptor antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-IL-4 sR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL4R(3566)

General description

Interleukin-4 (IL-4) is a T cell derived chemokine that stimulates the Th2 mediated immune response and proliferation of B cells. The effects of IL-4 are mediated by two types of receptors, type I receptor consisting of IL-4Rα and common γ chain and type II receptor composed of IL-4Rα and IL-13Rα1. The signalling stimulated by IL-4R leads to activation of JAK/STAT6 and IRS-mediated PI3K/Akt pathway. Through these pathways, IL-4 is responsible for endocytic activity of macrophages, chemotaxis of leukocytes in response to inflammation, angiogenesis and regulation of nitric oxide metabolism in macrophages. Anti-tumor effects of IL-4R signaling have been reported in cancers of breast, liver and renal cells
Anti-Interleukin-4 soluble receptor specifically recognizes human cell surface IL-4R.

Specificity

The antibody will neutralize human cell surface IL-4 receptor mediated-bioactivity.

Immunogen

recombinant human IL-4 soluble receptor (IL-4 sR), expressed in Sf 21 insect cells.

Application

Anti-Interleukin-4 soluble receptor antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. For ELISA, a working concentration of 0.5-1.0 μg/ml is recommended. The antibody is suitable for neutralization reactions at a working concentration of 5-10 μg/ml. This antibody was used for immunostaining of parasympathetic neurons from guinea pig and human neuroblastoma cells at a dilution of 1:200.

Physical form

Lyophilized from 51.5 μL of a 0.2μm filitered solution in PBS (pH7.4) with 5% trehalose.

Preparation Note

Purified using Protein G.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allison D Fryer et al.
The Journal of clinical investigation, 116(1), 228-236 (2005-12-24)
Eosinophils cluster around airway nerves in patients with fatal asthma and in antigen-challenged animals. Activated eosinophils release major basic protein, which blocks inhibitory M2 muscarinic receptors (M2Rs) on nerves, increasing acetylcholine release and potentiating vagally mediated bronchoconstriction. We tested whether
M Stein et al.
The Journal of experimental medicine, 176(1), 287-292 (1992-07-01)
Expression of the macrophage mannose receptor is inhibited by interferon gamma (IFN-gamma), a T helper type 1 (Th-1)-derived lymphokine. Interleukin 4 (IL-4), a Th-2 lymphocyte product, upregulates major histocompatibility class II antigen expression but inhibits inflammatory cytokine production by macrophages.
C K Oh et al.
European respiratory review : an official journal of the European Respiratory Society, 19(115), 46-54 (2010-10-20)
Asthma is a complex, persistent, inflammatory disease characterised by airway hyperresponsiveness in association with airway inflammation. Studies suggest that regular use of high-dose inhaled corticosteroids and long-acting bronchodilators or omalizumab (a humanised monoclonal antibody that binds to immunoglobulin E and
E M Coccia et al.
International immunology, 12(7), 977-985 (2000-07-06)
NO is a labile radical involved in several immunological, antimicrobial and inflammatory processes. In macrophages, NO formation is catalyzed by the cytokine-inducible enzyme inducible NO synthase (iNOS). The importance of IFN regulatory factor (IRF)-1 and of the signal transducers and
Alberto Mantovani et al.
Trends in immunology, 25(12), 677-686 (2004-11-09)
Plasticity and functional polarization are hallmarks of the mononuclear phagocyte system. Here we review emerging key properties of different forms of macrophage activation and polarization (M1, M2a, M2b, M2c), which represent extremes of a continuum. In particular, recent evidence suggests

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service