Skip to Content
Merck
All Photos(6)

Key Documents

HPA029025

Sigma-Aldrich

Anti-INTS11 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CPSF3L, Anti-CPSF73L, Anti-FLJ20542, Anti-INT11, Anti-RC-68, Anti-INTS11, Anti-cleavage and polyadenylation specific factor 3-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LVGQAEPESVLLVHGEAKKMEFLKQKIEQELRVNCYMPANGETVTLPTSPSIPVGISLGLLKREMAQGLLPEAKKPRLLHGTLIMKDSNFRLVSSEQALK

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CPSF3L(54973)

General description

The gene CPSF3L (cleavage and polyadenylation-specific factor 3-like, also referred to as INTS11 (integrator complex subunit 11)) is mapped to human chromosome 1p36.33. The protein is present in the cytoplasm and nucleus. It forms a heterodimer with INTS9 and the dimer is responsible for snRNA (small nuclear RNA) 3′ end processing.

Immunogen

integrator complex subunit 11 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CPSF3L antibody produced in rabbit has been used in western blotting.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76720

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas Ferreira Dasilva et al.
Science advances, 7(45), eabe3393-eabe3393 (2021-11-04)
[Figure: see text].
A CPSF-73 homologue is required for cell cycle progression but not cell growth and interacts with a protein having features of CPSF-100.
Dominski Z, et al.
Molecular and Cellular Biology, 25, 1489-1500 (2005)
The Chemical Biology of Human Metallo-?-Lactamase Fold Proteins.
Pettinati I, et al.
Trends in Biochemical Sciences, 41, 338-355 (2016)
Shiri Levy et al.
Nucleic acids research, 44(4), 1813-1832 (2016-01-31)
Post-transcriptional control of mitochondrial gene expression, including the processing and generation of mature transcripts as well as their degradation, is a key regulatory step in gene expression in human mitochondria. Consequently, identification of the proteins responsible for RNA processing and
Jingyin Yue et al.
Genes & development, 31(17), 1809-1820 (2017-10-07)
Activating mutations in the mitogen-activated protein kinase (MAPK) cascade, also known as the RAS-MEK-extracellular signal-related kinase (ERK1/2) pathway, are an underlying cause of >70% of human cancers. While great strides have been made toward elucidating the cytoplasmic components of MAPK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service