Skip to Content
Merck
All Photos(4)

Key Documents

HPA025065

Sigma-Aldrich

Anti-GTF2E2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FE, Anti-NCRNA00196, Anti-TF2E2, Anti-TFIIE-B, Anti-general transcription factor IIE, polypeptide 2, β

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PSLLRERELFKKRALSTPVVEKRSASSESSSSSSKKKKTKVEHGGSSGSKQNSDHSNGSFNLKALSGSSGYKFGVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GTF2E2(2961)

General description

The gene GTF2E2 (general transcription factor IIE subunit 2) is mapped to human chromosome 8p12. The protein has a serine-rich region, a centrally located forkhead domain, a leucine repeat motif, a region similar to the bacterial σ factor subdomain 3, a bHLH (basic region-helix-loop-helix) motif and a bHL (basic region-helix-loop) sequence.

Immunogen

general transcription factor IIE, polypeptide 2, beta 34kDa recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene GTF2E2 (general transcription factor IIE subunit 2) encodes the β subunit of transcription factor IIE (TFIIEβ). It has DNA binding ability and is essential for transcription. GTF2E2 also binds to RNA polymerase II. In patients with trichothiodystrophy, mutation in GTF2E2 is responsible for destabilization of TFIIE.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76441

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoshu Bi et al.
Cancer cell international, 21(1), 181-181 (2021-03-25)
Lung adenocarcinoma (LUAD) is one of the most common malignancies worldwide. However, the molecular mechanism of LUAD tumorigenesis and development remains unclear. The purpose of this study was to comprehensively illustrate the role of GTF2E2 in the growth and progression
Christiane Kuschal et al.
American journal of human genetics, 98(4), 627-642 (2016-03-22)
The general transcription factor IIE (TFIIE) is essential for transcription initiation by RNA polymerase II (RNA pol II) via direct interaction with the basal transcription/DNA repair factor IIH (TFIIH). TFIIH harbors mutations in two rare genetic disorders, the cancer-prone xeroderma
Aki Tanaka et al.
Genes to cells : devoted to molecular & cellular mechanisms, 14(3), 395-405 (2009-02-13)
The human general transcription factor, TFIIE, consists of two subunits, alpha and beta. Structural analyses indicated the presence of a forkhead motif within the central region of TFIIEbeta. This motif was essential for transcription and possessed a double-stranded DNA-binding activity.
Angelica Ronald et al.
Behavior genetics, 40(1), 31-45 (2009-12-17)
Two separate genome-wide association studies were conducted to identify single nucleotide polymorphisms (SNPs) associated with social and nonsocial autistic-like traits. We predicted that we would find SNPs associated with social and non-social autistic-like traits and that different SNPs would be
Tomomichi Watanabe et al.
Molecular and cellular biology, 23(8), 2914-2926 (2003-04-01)
The general transcription factor TFIIE plays essential roles in both transcription initiation and the transition from initiation to elongation. Previously, we systematically deleted the structural motifs and characteristic sequences of the small subunit of human TFIIE (hTFIIE beta) to map

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service