Skip to Content
Merck
All Photos(3)

Key Documents

HPA024748

Sigma-Aldrich

Anti-CA8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CA-VIII, Anti-CARP, Anti-Carbonic anhydrase VIII, Anti-Carbonic anhydrase-related protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

IQYKGKSKTIPCFNPNTLLPDPLLRDYWVYEGSLTIPPCSEGVTWILFRYPLTISQLQIEEFRRLRTHVKGAELVEGCDGILGDNFRPTQP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CA8(767)

General description

The gene CA8 (carbonic anhydrase 8) is mapped to human chromosome 8q. It is an isozyme of α-carbonic anhydrases and is widely expressed. However, it is strongly expressed in the cerebellum. The protein does not exhibit catalytic carbonic anhydrase activity.

Immunogen

Carbonic anhydrase-related protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mutations in CA8 (carbonic anhydrase 8) gene causes neuropathology, such as ataxia, mild mental retardation and the predisposition to quadrupedal gait. It is also associated with the development of colorectal and lung cancers. Additionally, it is upregulated in various cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76406

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Namik Kaya et al.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics, 156B(7), 826-834 (2011-08-04)
We define the neurological characteristics of familial cases from multiple branches of a large consanguineous family with cerebellar ataxia, mental retardation (MR), and dysequilibrium syndrome type 3 caused by a mutation in the recently cloned CA8 gene. The linkage analysis
Makoto Nishikata et al.
Molecular carcinogenesis, 46(3), 208-214 (2007-01-16)
Increased expression of carbonic anhydrase-related protein (CA-RP) VIII has previously been shown in colorectal carcinoma. Since CA-RP has no catalytic carbonic anhydrase (CA) activity, the present study attempted to elucidate its biological significance in colon cancer cells. From a colon
Min-Syuan Huang et al.
Biochimica et biophysica acta, 1840(9), 2829-2842 (2014-05-06)
Carbonic anhydrase 8 (CA8) is an isozyme of α-carbonic anhydrases (CAs). Previous studies showed that CA8 can be detected in human adult brain, with more intense expression in the cerebellum. Single mutations in CA8 were reported to cause novel syndromes
Ashok Aspatwar et al.
Current pharmaceutical design, 16(29), 3264-3276 (2010-09-08)
Mammalian carbonic anhydrase (α-CA) gene family comprises sixteen isoforms, thirteen of which are active isozymes and three isoforms lack classical CA activity of reversible hydration of CO(2) due to absence of one or more histidine residues required for CA catalytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service