Skip to Content
Merck
All Photos(11)

Key Documents

HPA018842

Sigma-Aldrich

Anti-LIAS antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LS, Anti-Lip-syn, Anti-Lipoate synthase, Anti-Lipoic acid synthase, Anti-Lipoyl synthase, mitochondrial precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SKVRDPRANFDQSLRVLKHAKKVQPDVISKTSIMLGLGENDEQVYATMKALREADVDCLTLGQYMQPTRRHLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIAS(11019)

General description

The gene LIAS (lipoyl synthase - mitochondrial) is mapped to human chromosome 4p14. The protein is mainly present in the mitochondria.

Immunogen

Lipoyl synthase, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Lipoyl synthase - mitochondrial (LIAS) is involved in synthesis of lipoic acid, a mitochondrial antioxidant. Absence of LIAS leads to imbalance in the antioxidant defense system, thereby causing inflammation, insulin resistance and mitochondrial abnormalities. Mutations in LIAS are associated with nonketotic hyperglycinemia, characterized by deficiency in lipoylation of mitochondrial proteins. In addition, mutation is LIAS is linked with neonatal-onset epilepsy, muscular hypotonia, lactic acidosis and high glycine levels in plasma and urine.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74695

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter R Baker et al.
Brain : a journal of neurology, 137(Pt 2), 366-379 (2013-12-18)
Patients with nonketotic hyperglycinemia and deficient glycine cleavage enzyme activity, but without mutations in AMT, GLDC or GCSH, the genes encoding its constituent proteins, constitute a clinical group which we call 'variant nonketotic hyperglycinemia'. We hypothesize that in some patients
Masakatsu Hishizawa et al.
British journal of haematology, 130(3), 382-390 (2005-07-27)
Adult T-cell leukaemia (ATL) is a peripheral T-cell neoplasm caused by human T-cell leukaemia virus type I (HTLV-I). Several clinical observations suggest that some tumour-associated antigens in ATL may be recognised by the immune system. In this study, we performed
Indira Padmalayam et al.
Diabetes, 58(3), 600-608 (2008-12-17)
Lipoic acid synthase (LASY) is the enzyme that is involved in the endogenous synthesis of lipoic acid, a potent mitochondrial antioxidant. The aim of this study was to study the role of LASY in type 2 diabetes. We studied expression
NOTCH4I?L12_16 sensitizes lung adenocarcinomas to EGFR-TKIs through transcriptional down-regulation of HES1.
Zhang, et al.
Nature Communications, 14, 3183-3183 (2023)
Johannes A Mayr et al.
American journal of human genetics, 89(6), 792-797 (2011-12-14)
Lipoic acid is an essential prosthetic group of four mitochondrial enzymes involved in the oxidative decarboxylation of pyruvate, α-ketoglutarate, and branched chain amino acids and in the glycine cleavage. Lipoic acid is synthesized stepwise within mitochondria through a process that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service