Skip to Content
Merck
All Photos(3)

Key Documents

HPA018034

Sigma-Aldrich

Anti-SLC30A7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Solute carrier family 30 member 7, Anti-Zinc transporter 7, Anti-ZnT-7, Anti-Znt-like transporter 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

HGGHGHSHGSGHGHSHSLFNGALDQAHGHVDHCHSHEVKHGAAHSHDHAHGHGHFHSHDGPSLKETTGPSRQI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 30 member 7 (SLC30A7) belongs to the zinc transporter family SLC30A and is expressed in the small intestine and liver. This 387-amino acid protein is localized to the Golgi apparatus. It contains six transmembrane domains. The gene encoding SLC30A7 is located on chromosome 1.

Immunogen

Zinc transporter 7 recombinant protein epitope signature tag (PrEST)

Application

Anti-SLC30A7 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Solute carrier family 30 member 7 (SLC30A7) is a zinc transporter which transports zinc from the cytoplasm to the Golgi apparatus. It may be involved in the activation of the phosphoinositide 3-kinase (PI3K) pathway. Knockdown of SLC30A7 in rat peritoneal mesothelial cells (RPMCs) show an increase in apoptosis. This transporter activates an alkaline phosphatase and is needed for the proper functioning of leukocytes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73274

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gastrointestinal factors influencing zinc absorption and homeostasis.
RJ Cousins
International Journal for Vitamin and Nutrition Research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal International de Vitaminologie et de Nutrition, 80(4-5), 243-248 (2010)
Xiuli Zhang et al.
Cellular signalling, 25(4), 999-1010 (2013-01-01)
Zinc is an essential micronutrient and cytoprotectant involved in many types of apoptosis. The zinc transporter family SLC30A (ZnTs) is an important factor in the regulation of zinc homeostasis; however, its function in apoptosis in peritoneal mesothelial cells (PMCs) remains
Qingqing Chu et al.
International journal of molecular medicine, 37(6), 1619-1626 (2016-04-29)
Previous studies have demonstrated that zinc (Zn) is an essential trace element which is involved in male reproduction. The zinc transporter (ZnT) family, SLC30a, is involved in the maintenance of Zn homeostasis and in mediating intracellular signaling events; however, relatively little
Catherine P Kirschke et al.
The Journal of biological chemistry, 278(6), 4096-4102 (2002-11-26)
ZnT7, a novel member of the zinc transporter (ZnT) family, was identified by searching the expressed sequence tag (EST) databases with the amino acid sequence of ZnT1. Like the other ZnT proteins, the protein (387 amino acids) predicted from this
Silke Overbeck et al.
Journal of leukocyte biology, 83(2), 368-380 (2007-11-01)
Intracellular zinc homeostasis is strictly regulated by zinc binding proteins and zinc transporters. In the present study, we quantified in a first global view the expression of all characterized human zinc exporters (hZnT-1-9) in different leukocyte subsets in response to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service