Skip to Content
Merck
All Photos(8)

Key Documents

HPA003909

Sigma-Aldrich

Anti-UBE2I antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SUMO-conjugating enzyme UBC9, Anti-SUMO-protein ligase, Anti-Ubiquitin carrier protein 9, Anti-Ubiquitin carrier protein I, Anti-Ubiquitin-conjugating enzyme E2 I, Anti-Ubiquitin-protein ligase I, Anti-p18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

IALSRLAQERKAWRKDHPFGFVAVPTKNPDGTMNLMNWECAIPGKKGTPWEGGLFKLRMLFKDDYPSSPPKCKFEPPLFHPNVYPSGTVCLSILEEDKDWRPAITIKQILLGIQELLNEPNIQDPAQAEAYTIYCQNRVEYEKRVRAQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE2I(7329)

Immunogen

SUMO-conjugating enzyme UBC9 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SUMO-conjugating enzyme UBC9 is a protein encoded by the UBE2I gene in humans. It plays a key role in post-translation modification called sumoylation. It is considered as the sole conjugating enzyme for sumoylation that regulates protein function and plays an important role in tumorigenesis. Sumoylated proteins function in many cellular processes including cell growth, maintaining the genome integrity and stability and cancer development. The protein is expressed at higher levels in primary lung cancer tissue and metastatic nodules as compared to premalignant and/or normal tissue. Up-regulation of this gene expression promotes migration and invasion. It may play an important role in cancer progression by promoting invasion and metastasis in lung cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86437

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Wozniak et al.
Pathology oncology research : POR, 20(1), 67-72 (2013-07-23)
UBC9 protein (E2-conjugating enzyme) plays a key role in post-translation modification named sumoylation. Proteins, which are sumoylated take part in many cellular processes including cell growth, maintaining the genome integrity and stability and cancer development. The aim of this study
Hui Li et al.
Oncology reports, 29(4), 1588-1594 (2013-02-06)
Lung cancer is the leading cause of cancer-related mortality worldwide. The mortality is high mainly due to the lack of known effective screening procedures; there is a high tendency for early spread and systemic therapies do not cure metastatic disease.
Shi-Feng Chen et al.
Chinese journal of cancer, 30(9), 638-644 (2011-09-02)
Ubiquitin-conjugating enzyme 9 (Ubc9), the sole conjugating enzyme for sumoylation, regulates protein function and plays an important role in tumorigenesis. Whether Ubc9 is involved in the chemoresistance of breast cancer remains unknown. In this study, we aimed to evaluate the
A B Steffensen et al.
Acta physiologica (Oxford, England), 222(3) (2017-09-10)
The voltage-gated potassium channel KV 11.1 is the molecular basis for the IKr current, which plays an important role in cardiac physiology. Its malfunction is associated with both inherited and acquired cardiac arrhythmias. Native currents differ from those in experimental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service