Skip to Content
Merck
All Photos(2)

Key Documents

HPA002913

Sigma-Aldrich

Anti-GPM6B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-M6b antibody produced in rabbit, Anti-Neuronal membrane glycoprotein M6-b antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

AVPVFMFYNIWSTCEVIKSPQTNGTTGVEQICVDIRQYGIIPWNAFPGKICGSALENICNTNEFYMSYH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPM6B(2824)

General description

Neuronal membrane glycoprotein M6-b is a protein encoded by the GPM6B gene in humans and is located at Xp22.2. The protein belongs to the proteolipid protein family.

Immunogen

Neuronal membrane glycoprotein M6-b recombinant protein epitope signature tag (PrEST)

Application

Anti-GPM6B antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Neuronal membrane glycoprotein M6-b is a protein encoded by the GPM6B gene in humans and is located at Xp22.2. The protein belongs to the proteolipid protein family. It plays an important role in neural development. Defect in this gene is associated with several neurological disorders such as Aicardi syndrome, Rett syndrome, X-linked Charcot-Marie-Tooth neuropathy and X-linked mental retardation syndromes. The gene is up-regulated during osteoblast differentiation. It is a novel regulator of osteoblast function and bone formation. The gene is considered as a good candidate gene for Pelizaeus-Merzbacher-like syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74334

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinya Kusumoto et al.
Oncology letters, 25(2), 45-45 (2023-01-17)
Tumor cell plasticity and tumor heterogeneity are involved in therapy resistance. Cancer stem cells (CSCs) refer to tumor cells that have the ability to self-renew, and generate the diverse cells that comprise the tumor and complicate tumor heterogeneity. In recent
S Olinsky et al.
Genomics, 33(3), 532-536 (1996-05-01)
M6 is a neuronal membrane glycoprotein that may have an important role in neural development. This molecule was initially defined by a monoclonal antibody that affected the survival of cultured cerebellar neurons and the outgrowth of neurites. The nature of
Marco Henneke et al.
American journal of medical genetics. Part A, 128A(2), 156-158 (2004-06-24)
"Pelizaeus-Merzbacher-like syndrome" is an undetermined leukodystrophy disorder of diffuse hypomyelination. The patients' clinical phenotype is indistinguishable from classical Pelizaeus-Merzbacher disease (PMD), but the patients lack PLP1 gene duplications or mutations. They represent about 20% of all cases with a clinical
V Narayanan et al.
American journal of medical genetics, 78(2), 165-168 (1998-07-23)
Human Xp22.2 has been proposed as a candidate region for the Rett syndrome (RTT) gene. M6b, a member of the proteolipid protein gene family, was mapped to Xp22.2 within one of the RTT candidate regions. In this article we describe
Ksenija Drabek et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 26(9), 2045-2051 (2011-06-04)
Neuronal membrane glycoprotein gene (GPM6B) encodes a membrane glycoprotein that belongs to the proteolipid protein family. We identified GPM6B as a gene that is strongly upregulated during osteoblast differentiation. To investigate the role of GPM6B in the process of bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service