Skip to Content
Merck
All Photos(5)

Documents

H6279

Sigma-Aldrich

Heparin ammonium salt from porcine intestinal mucosa

≥140 USP units/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Porcine intestinal mucosa

Quality Level

form

powder

specific activity

≥140 USP units/mg

color

white to light yellow

solubility

water: 100 mg/mL, clear to hazy, faintly yellow to brownish-yellow

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arleta Krystyna Skrzynska et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 215, 35-44 (2017-10-29)
The influence of chronic stress, induced by food deprivation (FD) and/or high stocking density (HSD), was assessed on stress, vasotocinergic and isotocinergic pathways of the gilthead sea bream (Sparus aurata). Fish were randomly assigned to one of the following treatments:
E De la Roca et al.
Physiology & behavior, 173, 272-278 (2017-02-28)
To gain further insights on the wedge sole (Dicologoglossa cuneata Moreau, 1881) physiology and behavior, we evaluated its daily feeding and locomotor activity rhythms and compared three different feeding strategies: self-feeding (SF), diurnal feeding schedule (DS) and nocturnal feeding schedule
Mingjuan Wu et al.
The British journal of nutrition, 120(3), 269-282 (2018-05-26)
An 8-week growth trial was conducted to evaluate the effects of dietary arginine (Arg) levels on growth, gut morphology, oxidation resistance and immunity of hybrid grouper (Epinephelus fuscoguttatus♀×Epinephelus lanceolatus♂) juveniles. Seven isoenergetic (1465 kJ (350 kcal)/100-g DM), isoproteic (53·5 %
Zhiyu Zhou et al.
The British journal of nutrition, 125(4), 408-419 (2020-07-28)
A 6-week growth trial was conducted to evaluate the influences of dietary valine (Val) levels on growth, protein utilisation, immunity, antioxidant status and gut micromorphology of juvenile hybrid groupers. Seven isoenergetic, isoproteic and isolipidic diets were formulated to contain graded
Marcelino Herrera et al.
Animals : an open access journal from MDPI, 9(6) (2019-06-15)
: The objective of this study was to assess the impact of different dietary Trp concentrations on the stress and metabolism response of juvenile Senegalese soles (Solea senegalensis). Fish (38.1 ± 1.9 g) were fed different Trp-enriched feeds (0%, 1%

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service