Skip to Content
Merck
All Photos(1)

Key Documents

GW22458A

Sigma-Aldrich

Anti-LDLR antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

manufacturer/tradename

Genway 15-288-22458A

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LDLR(3949)

Immunogen

Immunogen Sequence: GI # 4504975, sequence 29-205
Recombinant low density lipoprotein receptor

Application

Anti-LDLR antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

The Low-Density Lipoprotein (LDL) Receptor is a mosaic protein encoded by the LDLR gene in humans and belongs to the low density lipoprotein receptor gene family. It is located on chromosome number 19. LDL receptor is a mosaic protein constituiting of exons shared with different proteins. It is crucial for maintenance of cholesterol homeostasis. A deficiency in LDLR leads to hypercholesterolemia. LDLR is a type I transmembrane protein that needs O-glycosylation for stable expression at the cell surface. Mutation in this gene may cause familial hypercholesterolemia (FH), an autosomal dominant disease.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulo Caleb Junior Lima Santos et al.
Atherosclerosis, 233(1), 206-210 (2014-02-18)
Familial hypercholesterolemia (FH) is an autosomal dominant disease caused mainly by LDLR mutations. This study assessed the influence of the presence and type of LDLR mutation on lipid profile and the response to lipid-lowering therapy in Brazilian patients with heterozygous
Isabel De Castro-Orós et al.
BMC medical genomics, 7, 17-17 (2014-04-09)
GWAS have consistently revealed that LDLR locus variability influences LDL-cholesterol in general population. Severe LDLR mutations are responsible for familial hypercholesterolemia (FH). However, most primary hypercholesterolemias are polygenic diseases. Although Cis-regulatory regions might be the cause of LDL-cholesterol variability; an
Jovan Nikolic et al.
Nature communications, 9(1), 1029-1029 (2018-03-14)
Vesicular stomatitis virus (VSV) is an oncolytic rhabdovirus and its glycoprotein G is widely used to pseudotype other viruses for gene therapy. Low-density lipoprotein receptor (LDL-R) serves as a major entry receptor for VSV. Here we report two crystal structures
Nis Borbye Pedersen et al.
The Journal of biological chemistry, 289(25), 17312-17324 (2014-05-07)
The low density lipoprotein receptor (LDLR) is crucial for cholesterol homeostasis and deficiency in LDLR functions cause hypercholesterolemia. LDLR is a type I transmembrane protein that requires O-glycosylation for stable expression at the cell surface. It has previously been suggested
U Francke et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(9), 2826-2830 (1984-05-01)
The availability of a species-specific monoclonal antibody that recognizes the low density lipoprotein (LDL) receptor of human but not hamster origin permitted assignment of the structural gene for the human receptor to chromosome 19. The antibody was used to detect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service