GE30152105011150
Sera-Mag™ Magnetic Streptavidin Coated particles
Cytiva 30152105011150, 1 μm avg. part. size, suspension (1% Solids)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115700
NACRES:
NA.56
Recommended Products
ligand
(Olido(dt) particles)
form
suspension (1% Solids)
packaging
package of 1 mL
manufacturer/tradename
Cytiva 30152105011150
concentration
10 mg/mL
avg. part. size
1 μm
shipped in
wet ice
storage temp.
2-8°C
Related Categories
Application
Sera-Mag™ SpeedBeads and Sera-Mag™ Carboxylate-Modified Magnetic Particles typical application uses include sample preparation, proteomics, nucleic acid isolation, and immunoassay applications.
Features and Benefits
Sera-Mag™ SpeedBeads and Sera-Mag™ Carboxylate-Modified Magnetic Particles allow convenient covalent coupling of target molecules.
- Carboxylic groups on the surface permit easy covalent coupling to biomolecules of interest using convenient carbodiimide chemistry.
- Different levels of hydrophobicity/hydrophilicity available.
- Available in Sera-Mag™ SpeedBeads and original Sera-Mag™ versions.
- Excellent sensitivity and low nonspecific binding for greater accuracy.
- Fast Magnetic Response Times and Stable Physical Integrity
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Sera-Mag is a trademark of Cytiva
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service