GE28-9322-93
Vivaspin 6 centrifugal concentrators
polyethersulfone membrane
Synonym(s):
Centrifugal concentrator, Lab concentrator, Sample concentrator
About This Item
Recommended Products
material
polycarbonate housing
polyethersulfone membrane
sterility
non-sterile
feature
wetted part: no
packaging
pkg of 25 ea
manufacturer/tradename
Cytiva 28932293
tube size
15 mL
working volume
6 mL
pore size
3 kDa MWCO
General description
Features and Benefits
- Ease-of-use: One-step sample concentration in a single tube, for minimal sample handling and reduced sample loss, as well as pH compatibility over a broad range – pH 1 to 9.
- Sample protection: Patented Vivaspin dead-stop technology ensures samples cannot be concentrated to dryness and enables direct concentrate recovery.
- Ease-of-use: One-step sample concentration in a single tube, for minimal sample handling and reduced sample loss.
- High flow rates: And minimal membrane blockage through the vertical polyethersulfone (PES) membrane design.
- Minimal protein adsorption: Thanks to the Vivaspin™ polycarbonate tube design.
- Wide selection: Vivaspin sample concentrators are suitable for sample volumes ranging from 100 μL to 20 mL, with a range of molecular weight cut-off (MWCO) values from Mr 3000 to 100 000.
Storage and Stability
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
This page describes troubleshooting strategies in purification of GST-tagged proteins using Cytiva products.
Protocols
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service