Skip to Content
Merck
All Photos(1)

Key Documents

GE10600001

Amersham Protran® Western blotting membranes, nitrocellulose

pore size 0.2 μm, roll W × L 300 mm × 4 m, pkg of 1 ea

Synonym(s):

Western blotting membrane, 0.2 NC nitrocellulose membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22
Pricing and availability is not currently available.

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600001

roll W × L

300 mm × 4 m

pore size

0.2 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

Amersham Protran® Western blotting membranes, nitrocellulose with a 0.2 μm pore size, offer low background and excellent signal-to-noise ratios for blotting protocols.

Application

Amersham Protran® Western blotting membranes, nitrocellulose is suitable for use in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Offers excellent signal-to-noise ratios, without the need for rigorous washing conditions
  • Compatible with various western blot labeling and detection systems
  • Decreased risk of “blow-through” by the high binding capacity of small proteins (< Mr 20 000)
  • Designed to detect chemiluminescence and fluorescence.
  • Proteins bound to the transfer membrane have a long shelf life
  • Precut sizes are available for western blot transfer applications
  • Provides excellent sensitivity, resolution, and low background specifically when used with Amersham ECL, Amersham ECL prime, or Amersham ECL select western blotting detection reagents

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
ECL is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Storage Class Code

4.1B - Flammable solid hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa Pascual et al.
Science advances, 6(20), eaax3868-eaax3868 (2020-05-23)
Organogenesis is directed by coordinated cell proliferation and differentiation programs. The hierarchical networks of transcription factors driving mammary gland development and function have been widely studied. However, the contribution of posttranscriptional gene expression reprogramming remains largely unexplored. The 3' untranslated
Bruno Pinto et al.
Neuron, 108(5), 887-904 (2020-10-08)
Microglia are brain-resident immune cells and regulate mechanisms essential for cognitive functions. Down syndrome (DS), the most frequent cause of genetic intellectual disability, is caused by a supernumerary chromosome 21, containing also genes related to the immune system. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service