Key Documents
GBL664293
Grace Bio-Labs silicone isolator sheet material
thickness 1.6 mm, black silicone sheet, no PSA
Synonym(s):
non-adhesive isolator, silicone blocker
About This Item
UNSPSC Code:
41122600
NACRES:
NB.17
Pricing and availability is not currently available.
material
red silicon
feature
black silicone sheet, no PSA
packaging
pack of 5
manufacturer/tradename
Grace Bio-Labs 664293
L × W
13 cm × 18 cm
180 mm × 130 mm
thickness
1.6 mm
General description
Silicone isolator sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended.Silicone sheet material is also available in CultureWell “water-clear” silicone, which is highly recommended for sensitive cell culture applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)