F3377
N-Formyl-L-methionine
≥90% (TLC)
Synonym(s):
fMet
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H11NO3S
CAS Number:
Molecular Weight:
177.22
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
N-Formyl-L-methionine, ≥90% (TLC)
Assay
≥90% (TLC)
form
powder
color
white
storage temp.
−20°C
SMILES string
CSCC[C@H](NC=O)C(O)=O
InChI
1S/C6H11NO3S/c1-11-3-2-5(6(9)10)7-4-8/h4-5H,2-3H2,1H3,(H,7,8)(H,9,10)/t5-/m0/s1
InChI key
PYUSHNKNPOHWEZ-YFKPBYRVSA-N
Application
N-Formyl-L-methionine (fMet) is used to identify, differentiate and characterize amino acid N-deformylase(s), N-carbamoylase(s) and N-aminoacylase(s).
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Tomsic et al.
The EMBO journal, 19(9), 2127-2136 (2000-05-03)
Binding of the 50S ribosomal subunit to the 30S initiation complex and the subsequent transition from the initiation to the elongation phase up to the synthesis of the first peptide bond represent crucial steps in the translation pathway. The reactions
Dongli Pan et al.
Molecular cell, 25(4), 519-529 (2007-02-24)
Translocation requires large-scale movements of ribosome-bound tRNAs. Using tRNAs that are proflavin labeled and single-turnover rapid kinetics assays, we identify one or possibly two kinetically competent intermediates in translocation. EF-G.GTP binding to the pretranslocation (PRE) complex and GTP hydrolysis are
Rouven Bingel-Erlenmeyer et al.
Nature, 452(7183), 108-111 (2008-02-22)
Messenger-RNA-directed protein synthesis is accomplished by the ribosome. In eubacteria, this complex process is initiated by a specialized transfer RNA charged with formylmethionine (tRNA(fMet)). The amino-terminal formylated methionine of all bacterial nascent polypeptides blocks the reactive amino group to prevent
M Guenneugues et al.
The EMBO journal, 19(19), 5233-5240 (2000-10-03)
The interaction between fMet-tRNA(f)(Met) and Bacillus stearothermophilus translation initiation factor IF2 has been characterized. We demonstrate that essentially all thermodynamic determinants governing the stability and the specificity of this interaction are localized within the acceptor hexanucleotide fMet-3'ACCAAC of the initiator
Tetsuo Ohmachi et al.
Applied microbiology and biotechnology, 65(6), 686-693 (2004-08-10)
N-carbamoyl-L-cysteine amidohydrolase (NCC amidohydrolase) was purified and characterized from the crude extract of Escherichia coli in which the gene for NCC amidohydrolase of Pseudomonas sp. strain ON-4a was expressed. The enzyme was purified 58-fold to homogeneity with a yield of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service