EP3125000141
Eppendorf Research® Plus Move It pipette
no. of channels, 12, volume 0.5-100 μL, 4.5-9 mm cone distance, light yellow
Synonym(s):
Eppendorf® Research® Plus Move It pipette, Pipettor, multichannel pipette, multichannel pipettor
About This Item
Recommended Products
material
light yellow
feature
adjustable
manual
no. of channels 12
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 3125000141
volume
0.5-100 μL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- 4-, 6-, 8- and 12-channel air-cushion pipettes with variable volume and adjustable tip spacing option
- Available as mechanical (Research plus) and electronic (Xplorer plus) adjustable tip spacing multi-channel pipette
- For plates (384-, 96-, 48-, 24- and 12-well), tubes, agarose gels and more
- Tip spacing freely selectable from 4.5 to 33 mm, depending on pipette variant
- Format limiter for quick switches backwards and forwards between source and destination formats
- Manual adjustment knob for smooth vibration-free format switches and liquid transfers without dripping
- Tubeless system for enhanced reliability and safety
- Autoclavable for user and sample safety (Research plus fully, and Xplorer plus lower part only) 360° rotatable pipette head for comfortable readability of display and fast identification of parameters
- Optimally balanced pipette - reduces hand fatigue for stress-free operation
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service