Skip to Content
Merck
All Photos(1)

Key Documents

EP0030108523

Eppendorf® DNA LoBind tubes

new

capacity 25 mL, PCR clean, cap (snapTec), withstand 17,000xg, pkg of 200 ea (5 bags x 40 tubes)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

material

cap (snapTec)
conical bottom
polypropylene

sterility

non-sterile

feature

DNase free
PCR clean
RNase free
graduations: no
lid

packaging

pkg of 200 ea (5 bags x 40 tubes)

manufacturer/tradename

Eppendorf® 0030108523

parameter

17,000 x g rpm speed

technique(s)

DNA purification: suitable
PCR: suitable

capacity

25 mL

color

colorless

suitability

suitable for PCR
suitable for molecular biology

binding type

DNA low binding

Looking for similar products? Visit Product Comparison Guide

General description

DNA LoBind Tubes, SnapTec cap, DNA LoBind, 25 mL, conical tubes, PCR clean, colorless, 200 tubes (5 bags × 40 tubes)

Features and Benefits

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Precise lid sealing for minimal evaporation rates in tube format
  • Rated up to 17,000_x_g centrifugation speed for molecular biology applications

Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service