Skip to Content
Merck
All Photos(1)

Key Documents

EMU022211

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Scyl1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCCAGCTGAGAAGCAGAAGTTCTTCCAAGAGCTGAGCAAGAGTCTAGACTCATTTCCCGAAGATTTCTGTCGACACAAGGTGCTGCCCCAGCTACTGACTGCCTTTGAGTTTGGCAATGCTGGGGCCGTGGTCCTCACACCTCTCTTCAAGGTGGGAAAATCCCTCCGTGCTGAAGAGTACCAGGAGAAGATCATCCCTGTGGTAGTTAAGATGTTCTCATCCACCGACCGGGCCATGCGCATCCGCCTCCTCCAGCAGATGGAGCAGTTCATCCAATACCTTGATGAGCCAACAGTCAACACGCAGATTTTCCCCCACGTCACACATGGCTTCCTGGACACCAACCCCGCCATCCGCGAGCAGACGGTCAAGTCCATGCTGCTCTTGGCCCCAAAGCTGAATGAGGCCAATCTCAATGTGGAACTGATGAAGCACTTTGCAAGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Hannus et al.
Nucleic acids research, 42(12), 8049-8061 (2014-05-31)
Short interfering RNAs (siRNAs) are widely used as tool for gene inactivation in basic research and therapeutic applications. One of the major shortcomings of siRNA experiments are sequence-specific off-target effects. Such effects are largely unpredictable because siRNAs can affect partially
Carsten Riether et al.
Cell reports, 34(4), 108663-108663 (2021-01-28)
Self-renewal is a key characteristic of leukemia stem cells (LSCs) responsible for the development and maintenance of leukemia. In this study, we identify CD93 as an important regulator of self-renewal and proliferation of murine and human LSCs, but not hematopoietic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service