The original Duolink sequences are linked to the Red color in the Multicolor detection kit.
DUO96000
Duolink® PLA Multicolor Reagent Pack
Synonym(s):
in situ Proximity Ligation Assay Kit, Protein Protein Interaction Kit
About This Item
Recommended Products
product line
Duolink®
technique(s)
immunofluorescence: suitable
multiplexing: suitable
proximity ligation assay: suitable
suitability
suitable for fluorescence
shipped in
dry ice
storage temp.
−20°C
Application
Fluorescence detection reagents are often used with FITC, Cy®3,Texas Red® or Cy®5 filter. The filter chosen is dependent on the color of the Multicolor Probemaker kit selected.
Application Note
Primary antibodies are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC), or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Duolink® in situ reagents are suitable for use on fixed cells, cytospin cells, cells grown on slide, formalin-fixed, paraffin embedded (FFPE), or tissue (fresh or frozen). No minimum number of cells is required.
Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects
View full Duolink® product list
Features and Benefits
- Visualize and detect multiple proteins events in one experiment.
- No overexpression or genetic manipulation required
- High specificity (fewer false positives)
- Detection of low abundant proteins due to rolling circle amplification
- Relative quantification possible
- No special equipment needed
- Faster and more simple than FRET
- Increased accuracy compared to co-IP
- Publication-ready results
Components
Duolink® PLA Multicolor Reagent Pack contains all the necessary reagents to perform the amplification and detection of bound custom PLA probes generated using Duolink® PLA Multicolor Probemaker Kits. Sufficient components are provided for the indicated number of reactions (30 or 100 RXN), based on 40 mL of the total reaction mixture covering 1 cm2.
- This product is comprised of the following:
- 5X Multicolor PLA LIgation Buffer DUO86002
- Ligase DUO82027 or DUO82025
- 5X Amplification Buffer DUO82050
- Polymerase DUO82028 or DUO82026
- 5X Multicolor PLA Detection Buffer DUO86001
- 1X Blocking Buffer DUO82007
- 1X Probemaker PLA Probe Diluent DUO82036
See datasheet for more information.
To perform a Multicolor Duolink® PLA experiment you will need primary antibodies (PLA, IHC, ICC or IF validated) that recognize target epitopes. You will also need at least two pair of the resulting Duolink® PLA Multicolor Probemaker-generated probes (Red A/Red B, Green C/Green D, Orange F/Orange G, and/or FarRed H/FarRed I).The reagents in the Duolink® PLA Multicolor Reagent Pack are required to perform the amplification and detection of bound Multicolor PLA probes. Recommended additional reagents include Duolink® Wash Buffers and Mounting Medium.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Duolink® PLA Multicolor technology and the quantification of protein-protein interactions in fixed tissues and cells.
Protocols
Duolink® PLA Multicolor Detection Protocol
-
Can the detection kit be used with the original Duolink sequences (DUO92002, rabbit plus & DUO92004, Mouse minus)?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service