Skip to Content
Merck
All Photos(1)

Key Documents

CLS480143

Corning® LSE rotors for LSE compact centrifuges

combination rotor for 6 x 15 mL, 2 x 50 mL and 2 x 15 mL

Synonym(s):

fixed angle centrifuge rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.13
Pricing and availability is not currently available.

feature

to hold 6 x 15 mL tubes
to hold 6 x 5 mL microcentrifuge tubes
to hold 6 x 50 mL tubes

packaging

case of 1
pack of 1

manufacturer/tradename

Corning 480143

parameter

200-6000 max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models LSE Compact

Looking for similar products? Visit Product Comparison Guide

General description

Combination rotor is an innovative multi-purpose rotor designed by Corning to add more versatility. This rotor can hold the various tube sizes including a 5 mL microcentrifuge tube, as well as round and conical bottom 15 mL and 50 mL centrifuge tubes. Rotor holds tubes at an increased fixed angle to advance the efficiency of cellular pelleting. An equal g-force is applied to the samples when spinning 15 mL and 50 mL tubes simultaneously.

Legal Information

Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service